Z238074
Monogoggles
green Frame
About This Item
Código UNSPSC:
46181802
Productos recomendados
disponibilidad
not available in EU
color
green Frame
Descripción general
Exclusive two-piece frame design. A separate nose piece of soft, pliable plastic molds to fit a variety of facial contours, providing better contact and a tighter seal around the nose and cheeks. Lightweight, transparent frames have an indirect ventilation system built-in to create an airflow across the lens, minimizing condensation. Lenses are made from clear, 0.060 in. thick, optical grade PC and are coated with FogGard to reduce fogging. Padded felt frames improve fit for workers with smaller facial features. Meets ANSI Z87.1 and CSA Z94.3 requirements. SEI certified.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)