Z236896
Gas-cylinder bracket
Double, bench-mount
About This Item
Código UNSPSC:
41100000
NACRES:
NB.51
Al × An × P
4 1/4 in. × 24 in. × 4 1/4 in.
peso
10.7 lb
Descripción general
Durable supports hold gas, oxygen, welding, cascade, propane, or LP cylinders, with diameters 3 to 12 3/8 in. Heavy 11 gauge welded construction, baked enamel finish, steel-reinforced resin edge-guards. 1 1/2 in.
Support straps are 1.5 in. wide by 54 in. long polypropylene with steel cinch buckle and rate a robust 1138 PSI and are the primary means of support as they enable the cylinders to be held tight and secure against the brackets, chain sets assure long service life. The bench mount brackets are secured via thumbscrews. All units comply with OSHA, NFPA, UFC, and AGA standards.
Support straps are 1.5 in. wide by 54 in. long polypropylene with steel cinch buckle and rate a robust 1138 PSI and are the primary means of support as they enable the cylinders to be held tight and secure against the brackets, chain sets assure long service life. The bench mount brackets are secured via thumbscrews. All units comply with OSHA, NFPA, UFC, and AGA standards.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)