Documentos
Z203335
Sigma-Aldrich
Aldrich® vacuum-transfer adapter
joint: ST/NS 29/32
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
joint
joint: ST/NS29/32
stopcock size
4 mm
General description
PTFE stopcocks. For transfer of hazardous and sensitive reagents.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC