Key Documents
Z151696
Sigma-Aldrich
Polished IR crystal window
KBr
About This Item
UNSPSC Code:
12000000
NACRES:
SB.32
material
KBr
technique(s)
IR spectroscopy: suitable
diam. × thickness
13 mm × 2 mm
General description
Use with magnetic holder (Z112488) for films, neat, or Nujol samples.