Merck
Z126608
Sigma-Aldrich
Giant-size molecular model kit
standard kit
Seleccione un Tamaño
1 KIT
US$ 751,00
US$ 751,00
PRECIOS SIN IMPUESTOS NACIONALES
Check Cart for Availability
Seleccione un Tamaño
Cambiar Vistas
1 KIT
US$ 751,00
About This Item
Código UNSPSC:
60104506
NACRES:
NB.17
US$ 751,00
PRECIOS SIN IMPUESTOS NACIONALES
Check Cart for Availability
descripción
no. of flexible bonds 8
no. of rigid bonds 104
Características
no. of atoms 50
Descripción general
Large, lightweight molecular models for classroom demonstration of organic and inorganic structures. For building models to scale of 20 cm/Å and 8 cm/Å. Bond lengths range from 5.5 to 27 cm. Atomic centers are 5 cm in diameter, finished in standard atom colors, and have a reinforced vinyl receptor at each bond position for insertion of connecting rods (bonds). All model kits include storage box and instructions. Additional atoms and bonds are available separately.
Características y beneficios
Includes 12 assorted plastic orbital connectors.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)
Visite la Librería de documentos
Los clientes también vieron
Questions
  1. What is the difference between these three products Z231312, Z231320, Z126608? There is no much information on the website.
    1 answer
    1. The difference between all three items is the number of molecules and bonds in each kit. Below are dimensions and the number of bonds and molecules are in each kit:
      Z126608
      • no. of flexible bonds 8
      • no. of rigid bonds 104
      • no. of atoms 50
      Z231320
      • no. of flexible bonds 32
      • no. of rigid bonds 100
      • no. of atoms 64
      Z231312
      • no. of flexible bonds 16
      • no. of rigid bonds 50
      • no. of atoms 32