Documentos
Z125539
Ace-Thred Thermometer/Tubing Adapter
With hose connection, joint: ST/NS 24/40, thread I.D. 7.0 mm
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
label
F
joint
joint: ST/NS24/40
manufacturer/tradename
Ace Glass 526116
thread I.D.
7.0 mm
General description
Threaded nylon bushing with FETFE® O-ring.
Legal Information
Ace-Thred is a trademark of Ace Glass, Inc.
FETFE is a registered trademark of Ace Glass, Inc.
replacement part