Documentos clave
Y906492
Sigma-Aldrich
Valve
brass bellows, 1/4 in. NPT straight path, external
About This Item
Código UNSPSC:
12142200
NACRES:
NA.12
Note: This product is only available with the purchase of a bulk gas product and not for individual sale. For more information, please contact Stable Isotopes Customer Service
Materiales
brass bellows
Parámetros
1000 psi max. pressure
Longitud
1.9375 in.
peso
159 g
Adaptador
1/4 in. NPT straight path, external
Categorías relacionadas
Otras notas
This product is only available with the purchase of a bulk gas product and not for individual sale. For more information, please contact Stable Isotopes Customer Service
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)