Merck
DWK92812F-100
KIMBLE® KIMAX® Heavy duty volumetric wide-mouth flask with PTFE stopper
glass flask, flask capacity (100 mL), class A
Seleccione un Tamaño
6 EA
US$ 477,00
US$ 477,00
PRECIOS SIN IMPUESTOS NACIONALES
Check Cart for Availability
Seleccione un Tamaño
Cambiar Vistas
6 EA
US$ 477,00
About This Item
Código UNSPSC:
41121804
NACRES:
NB.43
US$ 477,00
PRECIOS SIN IMPUESTOS NACIONALES
Check Cart for Availability
Materiales
PTFE stopper
glass flask
Características
class A
envase
case of 6 ea
fabricante / nombre comercial
Kimble® 92812F-100
matraz capacidad
(100 mL)
Descripción general
Class A Heavy Duty Wide Mouth Volumetric Flasks with PTFE Stopper
  • Pipet access is easy with wide-mouth volumetric flasks
  • Superior strength, durability and safety from heavy, uniform walls
  • Quick identification with large, permanent, easy-to-read markings
  • Flat interior bottoms for stirring with standard stir bars
  • Graduated to Class A volumetric tolerances
  • Standard Taper glass stoppers
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Características y beneficios
  • Manufactured from borosilicate glass 3.3
  • Wide mouth opening increases efficiency when performing large numbers of dilutions using volumetric pipets
  • Heavy duty walls provide superior strength, durability and safety
  • Wide mouth minimizes glass-to-glass contact allowing greater flexibility in hand movements which reduces the risk of spills
  • Wide mouth opening increases efficiency when performing large numbers of dilutions using volumetric pipets
  • Printed large volume numbers allow quick identification when selecting glassware
  • Flat interior bottoms for stirring with standard stir bars
  • Calibrate to contain
  • Graduated to Class A volumetric tolerances
Información legal
KIMBLE is a registered trademark of DWK Life Sciences
Kimax is a registered trademark of Gerresheimer Glass, Inc.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)