Documentos clave
CAD4851
Stainless steel 304 syringe needle
gauge 16, L 6 in.
Seleccione un Tamaño
12 EA
US$ 463,00
US$ 463,00
Fecha estimada de envío16 de abril de 2025
Seleccione un Tamaño
Cambiar Vistas
12 EA
US$ 463,00
About This Item
UNSPSC Code:
42142600
NACRES:
NB.03
US$ 463,00
Fecha estimada de envío16 de abril de 2025
description
Standard Hub with 304 Stainless Steel Tubing
packaging
pack of 12 ea
manufacturer/tradename
Cadence Science 4851
L
6 in.
gauge
16
General description
304 SS needle with chrome-plated brass Luer hub. Needle is designed with a slightly curved, deflecting tip to prevent coring during needle insertion through the septum.
Standard Hub with 304 Stainless Steel Tubing
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)