Documentos clave
BR4630350
BRAND® Dispensette® S Organic, Digital bottle-top dispenser
volume range 2.5-25 mL, without recirculation valve
Seleccione un Tamaño
1 EA
US$ 1.200,00
US$ 1.200,00
PRECIOS SIN IMPUESTOS NACIONALES
Check Cart for Availability
Seleccione un Tamaño
Cambiar Vistas
1 EA
US$ 1.200,00
About This Item
Código UNSPSC:
41121500
NACRES:
NB.01
US$ 1.200,00
PRECIOS SIN IMPUESTOS NACIONALES
Check Cart for Availability
Características
incremental 0.1 mL
DE-M marking
without recirculation valve
fabricante / nombre comercial
BRAND 4630350
intervalo de volumen
2.5-25 mL
Adaptador
(GL 32-33)
GL 38
S 40
Descripción general
Dispensette S Organic is ideal for dispensing of organic solvents and acids
Características y beneficios
  • Extremely easy filling and dispensing
  • Simple priming by optimized flow channels
  • Rapid and safe volume adjustment with internal scalloped track
Otras notas
Digital-The mechanical volume setting is easy to read and ensures accurate and reproducible volume control. Easy Calibration Technique for calibration adjustment in seconds without tools. A calibration flag indicates changes from factory settings.
Información legal
BRAND is a registered trademark of BRAND GMBH + CO KG
Dispensette is a registered trademark of BRAND GMBH + CO KG
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)
Visite la Librería de documentos
Los clientes también vieron
Protocolos
This article provides testing instructions for BRAND Dispensette® S bottle-top liquid dispenser according to the standard ISO DIS 8655.
Contenido relacionado
Chemical compatibility chart for all Dispensette® S and Dispensette® S Organic models.