Merck
81240
Sigma-Aldrich
Poli(etilenglicol)
average Mn 4,000, hydroxyl
Sinónimos:
PEG
About This Item
Fórmula lineal:
H(OCH2CH2)nOH
Número de CAS:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.23
product name
Poli(etilenglicol), average Mn 4,000, platelets
biological source
platelets
form
powder
mol wt
Mn 3,500-4,500
average Mn 4,000
mp
58-61 °C
Ω-end
hydroxyl
α-end
hydroxyl
SMILES string
C(CO)O
InChI
1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2
InChI key
LYCAIKOWRPUZTN-UHFFFAOYSA-N
Categorías relacionadas
General description
Polyethylene glycol (PEG) is a hydrophilic polymer. It can be easily synthesized by the anionic ring opening polymerization of ethylene oxide, into a range molecular weights and variety of end groups. When crosslinked into networks PEG can have high water content, forming “hydrogels PEG is a suitable material for biological applications because it does not trigger an immune response.
Application
Polyethylene glycol (PEG) platelets were used:
  • in PEG-directed gene delivery of protoplast transfection and incubation
  • to prepare extraction for measuring cytosolic phosphoenolpyruvate carboxykinase (PEPCK) from plant tissues
  • as DNA transformation agent in a study of transient gene expression of plant protoplasts, derived from rice leaf sheaths.
  • in preparing isothermal assembly buffer and TSS broth preparation for unique nucleotide sequence (UNS)-guided assembly of repetitive DNA parts
  • in transformation of DNA into log-phase cells of the budding yeast Saccharomyces cerevisiae.
  • as DNA transformation reagent
Other Notes
Polymer networks with grafted cell adhesion peptides for highly biospecific cell adhesive substrates
Storage Class
11 - Combustible Solids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Visite la Librería de documentos
Los clientes también vieron
A simple and effective method to encapsulate tobacco mesophyll protoplasts to maintain cell viability
Lei R, et al.
Methods, 24-32 (2015)
Jennifer DeMars Tripp et al.
Yeast (Chichester, England), 30(5), 191-200 (2013-03-14)
Chemical-based methods have been developed for transformation of DNA into log-phase cells of the budding yeast Saccharomyces cerevisiae with high efficiency. Transformation of early stationary-phase cells, e.g. cells grown in overnight liquid cultures or as colonies on plates, is less
Shen, Jinbo, et al. "Isolation, culture, and transient transformation of plant protoplasts
Shen J, et al.
Current Protocols in Cell Biology, 2014, 2-8 null
Oleg Raitskin et al.
PloS one, 14(2), e0211598-e0211598 (2019-02-28)
Molecular tools adapted from bacterial CRISPR (Clustered Regulatory Interspaced Short Palindromic Repeats) systems for adaptive immunity have become widely used for plant genome engineering, both to investigate gene functions and to engineer desirable traits. A number of different Cas (CRISPR-associated)
Toshihiko Takada et al.
Gut microbes, 11(6), 1662-1676 (2020-06-20)
Probiotic products have been shown to have beneficial effects on human hosts, but what happens in the gastrointestinal tract after its ingestion remains unclear. Our aim was to investigate the changes within the small intestines after a single intake of