Merck
265888
Sigma-Aldrich
Lead
wire, diam. 1.0 mm, 99.99% trace metals basis
Sinónimos:
Lead element
About This Item
Fórmula empírica (notación de Hill):
Pb
Número de CAS:
Peso molecular:
207.20
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23
assay
99.99% trace metals basis
form
wire
resistivity
20.65 μΩ-cm
diam.
1.0 mm
bp
1740 °C (lit.)
mp
327.4 °C (lit.)
SMILES string
[Pb]
InChI
1S/Pb
InChI key
WABPQHHGFIMREM-UHFFFAOYSA-N
Quantity
8.9 g = 100 cm; 44.5 g = 500 cm
pictograms
Health hazardEnvironment
signalword
Danger
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Lact. - Repr. 1A - STOT RE 1 Oral
target_organs
Central nervous system,Blood,Immune system,Kidney
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk_germany
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type P3 (EN 143) respirator cartridges
Visite la Librería de documentos
Los clientes también vieron
Slide 1 of 1
1 of 1
Images in clinical medicine. Pellets in the appendix.
Ibrahim Zardawi et al.
The New England journal of medicine, 369(6), e7-e7 (2013-08-09)
Kai-Ying Chiang et al.
Journal of environmental sciences (China), 25(3), 626-636 (2013-08-08)
This study investigates biogeochemical reductive release of arsenate from beudantite into solution in a crater area in northern Taiwan, using a combination of X-ray absorption near-edge structure (XANES) and atomic absorption spectrometry. Total arsenic (As) concentrations in the soil were
Karsten Witt et al.
Brain : a journal of neurology, 136(Pt 7), 2109-2119 (2013-06-27)
Deep brain stimulation of the subthalamic nucleus improves motor functions in patients suffering from advanced Parkinson's disease but in some patients, it is also associated with a mild decline in cognitive functioning about one standard deviation from the preoperative state.
Plinio Carta et al.
Giornale italiano di medicina del lavoro ed ergonomia, 35(1), 32-35 (2013-06-27)
In several recent epidemiological studies blood lead levels (BLLs) even below the current CDC intervention level of 10 microg/dl have been associated with reduced neurocognitive capacities of children, with no clear evidence of a "safe" threshold. We analyzed the relationship
Muhammad Jamal Haider et al.
Roczniki Panstwowego Zakladu Higieny, 64(1), 37-42 (2013-06-26)
OBJECTIVE. The present study was carried out to investigate the effects of occupational lead exposure on the hematological and biochemical parameters in occupationally exposed and non exposed inhabitants of Karachi. MATERIAL AND METHODS. In 100 lead exposed subjects recruited from