HPLC Analysis of Biogenic Amines on Ascentis® RP-Amide
![HPLC Analysis of Biogenic Amines on Ascentis® RP-Amide application for HPLC](/deepweb/assets/sigmaaldrich/product/images/192/042/e7b5a4d8-a6ea-451d-a006-60454a641f26/800/e7b5a4d8-a6ea-451d-a006-60454a641f26.jpg)
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Ascentis RP-Amide, 15 cm x 2.1 mm I.D., 5 μm particles (565305-U)
mobile phase
[A] 0.1% ammonium acetate in water (pH unadjusted); [B] acetonitrile
gradient
25 to 75% B in 10 min; held at 75% B for 5 min
flow rate
0.5 mL/min
column temp.
35 °C
detector
UV, 340 nm
injection
10 μL
Description
Analysis Note
Following pre-column derivatization of primary amino groups using o-phthaldialdehyde/thiol reagent, the Ascentis RP-Amide was used for the analysis of biogenic amines commonly found in wines. These include histamine, spermine, spermidine, tyramine, putrescine and cadaverine. Structures along with an optimized chromatogram are presented.
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany