Merck
  • CDYL suppresses epileptogenesis in mice through repression of axonal Nav1.6 sodium channel expression.
CDYL suppresses epileptogenesis in mice through repression of axonal Nav1.6 sodium channel expression.
Nature communications (2017-08-27)
Yongqing Liu, Shirong Lai, Weining Ma, Wei Ke, Chan Zhang, Shumeng Liu, Yu Zhang, Fei Pei, Shaoyi Li, Ming Yi, Yousheng Shu, Yongfeng Shang, Jing Liang, Zhuo Huang
ABSTRACT
Impairment of intrinsic plasticity is involved in a range of neurological disorders such as epilepsy. However, how intrinsic excitability is regulated is still not fully understood. Here we report that the epigenetic factor Chromodomain Y-like (CDYL) protein is a critical regulator of the initiation and maintenance of intrinsic neuroplasticity by regulating voltage-gated ion channels in mouse brains. CDYL binds to a regulatory element in the intron region of SCN8A and mainly recruits H3K27me3 activity for transcriptional repression of the gene. Knockdown of CDYL in hippocampal neurons results in augmented Nav1.6 currents, lower neuronal threshold, and increased seizure susceptibility, whereas transgenic mice over-expressing CDYL exhibit higher neuronal threshold and are less prone to epileptogenesis. Finally, examination of human brain tissues reveals decreased CDYL and increased SCN8A in the temporal lobe epilepsy group. Together, our findings indicate CDYL is a critical player for experience-dependent gene regulation in controlling intrinsic excitability.Alterations in intrinsic plasticity are important in epilepsy. Here the authors show that the epigenetic factor CDYL regulates the gene expression of the voltage gated sodium channel, Nav1.6, which contributes to seizures in a rat model of epilepsy.
Sigma-Aldrich
MISSION® esiRNA, targeting human EZH2
Pricing and availability is not currently available.
Sigma-Aldrich
MISSION® esiRNA, targeting human REST
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-G9a (BAT8) Antibody, from rabbit, purified by affinity chromatography
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys27) Antibody, Upstate®, from rabbit
Pricing and availability is not currently available.
Sigma-Aldrich
N-Methyl-D-aspartic acid, ≥98% (TLC), solid
Pricing and availability is not currently available.