Merck
  • Molecular signaling underlying bulleyaconitine A (BAA)-induced microglial expression of prodynorphin.
Molecular signaling underlying bulleyaconitine A (BAA)-induced microglial expression of prodynorphin.
Scientific reports (2017-03-23)
Teng-Fei Li, Hai-Yun Wu, Yi-Rui Wang, Xin-Yan Li, Yong-Xiang Wang
ABSTRACT
Bulleyaconitine (BAA) has been shown to possess antinociceptive activities by stimulation of dynorphin A release from spinal microglia. This study investigated its underlying signal transduction mechanisms. The data showed that (1) BAA treatment induced phosphorylation of CREB (rather than NF-κB) and prodynorphin expression in cultured primary microglia, and antiallodynia in neuropathy, which were totally inhibited by the CREB inhibitor KG-501; (2) BAA upregulated phosphorylation of p38 (but not ERK or JNK), and the p38 inhibitor SB203580 (but not ERK or JNK inhibitor) and p38β gene silencer siRNA/p38β (but not siRNA/p38α) completely blocked BAA-induced p38 phosphorylation and/or prodynorphin expression, and antiallodynia; (3) BAA stimulated cAMP production and PKA phosphorylation, and the adenylate cyclase inhibitor DDA and PKA inhibitor H-89 entirely antagonized BAA-induced prodynorphin expression and antiallodynia; (4) The Gs-protein inhibitor NF449 completely inhibited BAA-increased cAMP level, prodynorphin expression and antiallodynia, whereas the antagonists of noradrenergic, corticotrophin-releasing factor, A1 adenosine, formyl peptide, D1/D2 dopamine, and glucagon like-peptide-1 receptors failed to block BAA-induced antiallodynia. The data indicate that BAA-induced microglial expression of prodynorphin is mediated by activation of the cAMP-PKA-p38β-CREB signaling pathway, suggesting that its possible target is a Gs-protein-coupled receptor - "aconitine receptor", although the chemical identity is not illustrated.
Sigma-Aldrich
MISSION® esiRNA, targeting human MAPK14
Pricing and availability is not currently available.
Sigma-Aldrich
Naphthol AS-E phosphate, suitable for histology
Pricing and availability is not currently available.
Sigma-Aldrich
MISSION® esiRNA, targeting human SYP
Pricing and availability is not currently available.
Sigma-Aldrich
SP600125, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
N,N-Dimethyldodecylamine, 97%
Pricing and availability is not currently available.
Sigma-Aldrich
2′,5′-Dideoxyadenosine, ≥95% (HPLC), solid
Pricing and availability is not currently available.