Merck
  • Regioselective Synthesis of α-Fluorinated Cyclopentenones by Organocatalytic Difluorocyclopropanation and Fluorine-Directed and Fluorine-Activated Nazarov Cyclization.
Regioselective Synthesis of α-Fluorinated Cyclopentenones by Organocatalytic Difluorocyclopropanation and Fluorine-Directed and Fluorine-Activated Nazarov Cyclization.
Chemistry (Weinheim an der Bergstrasse, Germany) (2016-12-03)
Kohei Fuchibe, Ryo Takayama, Takaharu Yokoyama, Junji Ichikawa
ABSTRACT
Silyl dienol ethers, prepared from α,β-unsaturated ketones, underwent proton sponge-catalyzed difluorocyclopropanation with trimethylsilyl 2,2-difluoro-2-fluorosulfonylacetate in a regioselective manner, leading to 1,1-difluoro-2-siloxy-2-vinylcyclopropanes in good yields. The cyclopropanes thus obtained were in turn subjected to fluoride-ion-catalyzed ring opening to afford 1-fluorovinyl vinyl ketones (i.e., Nazarov precursors). Treatment of the precursors with Me
Sigma-Aldrich
Tetrabutylammonium difluorotriphenylsilicate, 97%
Pricing and availability is not currently available.