Merck
  • Sodium orthovanadate suppresses DNA damage-induced caspase activation and apoptosis by inactivating p53.
Sodium orthovanadate suppresses DNA damage-induced caspase activation and apoptosis by inactivating p53.
Cell death and differentiation (2005-09-03)
A Morita, J Zhu, N Suzuki, A Enomoto, Y Matsumoto, M Tomita, T Suzuki, K Ohtomo, Y Hosoi
ABSTRACT
We previously reported that p42/SETbeta is a substrate for caspase-7 in irradiated MOLT-4 cells, and that treating the cells with sodium orthovanadate (vanadate) inhibits p42/SETbeta's caspase-mediated cleavage. Here, we initially found that the inhibitory effect of vanadate was due to the suppression of caspase activation but not of caspase activity. Further investigations revealed that vanadate suppressed upstream of apoptotic events, such as the loss of mitochondrial membrane potential, the conformational change of Bax, and p53 transactivation, although the accumulation, total phosphorylation, and phosphorylation of six individual sites of p53 were not affected. Importantly, vanadate suppressed p53-dependent apoptosis, but not p53-independent apoptosis. Finally, gel-shift and chromatin immunoprecipitation assays conclusively demonstrated that vanadate inhibits the DNA-binding activity of p53. Vanadate is conventionally used as an inhibitor of protein tyrosine phosphatases (PTPs); however, we recommend that the influence of vanadate not only on PTPs but also on p53 be considered before using it.
Sigma-Aldrich
Anti-p53 Antibody, aa 211-220, clone240, clone PAb240, Chemicon®, from mouse
Sigma-Aldrich
Sodium orthovanadate, 99.98% trace metals basis
Sigma-Aldrich
Anti-Phosphotyrosine Antibody, clone 4G10®, clone 4G10®, Upstate®, from mouse
Sigma-Aldrich
Anti-Histone H1 Antibody, clone AE-4, clone AE-4, Upstate®, from mouse