Merck
  • Determination of spirocyclic tetronic/tetramic acid derivatives and neonicotinoid insecticides in fruits and vegetables by liquid chromatography and mass spectrometry after dispersive liquid-liquid microextraction.
Determination of spirocyclic tetronic/tetramic acid derivatives and neonicotinoid insecticides in fruits and vegetables by liquid chromatography and mass spectrometry after dispersive liquid-liquid microextraction.
Food chemistry (2016-02-28)
Marta Pastor-Belda, Isabel Garrido, Natalia Campillo, Pilar Viñas, Pilar Hellín, Pilar Flores, José Fenoll
ABSTRACT
Dispersive liquid-liquid microextraction was used to preconcentrate three spirocyclic tetronic/tetramic acid derivatives (spirotetramat, spiromesifen and spirodiclofen) and five neonicotinoid (thiamethoxam, chlotianidin, imidacloprid, acetamiprid and thiacloprid) insecticides previously extracted from fruit and vegetable matrices with acetonitrile. The organic enriched phase was evaporated, reconstituted in 25μL acetonitrile and analyzed by reversed-phase liquid chromatography with tandem mass spectrometry using a triple quadrupole in selected reaction monitoring mode. Enrichment factors in the 15-100 range were obtained. A matrix effect was observed, the detection limits varying between 0.025 and 0.5ngg(-1), depending on the compound and the sample matrix. The developed method was applied to the analysis of 25 samples corresponding to five different fruit and vegetable matrices. Only thiamethoxam was detected in a lemon sample at a concentration close to the quantification limit, and spiromesifen and spirotetramat at concentrations between 11.6 and 54.5ngg(-1).
Product Description
Supelco
Spirodiclofen, PESTANAL®, analytical standard
Supelco
Spiromesifen, PESTANAL®, analytical standard
Supelco
Clothianidin, PESTANAL®, analytical standard
Supelco
N-(6-Chloro-3-pyridylmethyl)-N-cyano-N-methylacetamidine, PESTANAL®, analytical standard
Supelco
Imidacloprid, PESTANAL®, analytical standard