Skip to Content
Merck
All Photos(1)

Key Documents

33599

Supelco

Spiromesifen

PESTANAL®, analytical standard

Synonym(s):

3-Mesityl-2-oxo-1-oxaspiro[4.4]non-3-en-4-yl) 3,3-dimethylbutyrate

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$279.00

$279.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$279.00

About This Item

Empirical Formula (Hill Notation):
C23H30O4
CAS Number:
Molecular Weight:
370.48
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$279.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

Cc1cc(C)c(c(C)c1)C2=C(OC(=O)CC(C)(C)C)C3(CCCC3)OC2=O

InChI

1S/C23H30O4/c1-14-11-15(2)18(16(3)12-14)19-20(26-17(24)13-22(4,5)6)23(27-21(19)25)9-7-8-10-23/h11-12H,7-10,13H2,1-6H3

InChI key

GOLXNESZZPUPJE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Spiromesifen belongs to the class of pesticides, which are derivatives of spirocyclic tetronic acid[1], used to control pests like whiteflies and mites.[2][3]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

The Whitefly, Bemisia tabaci (Homoptera: Aleyrodidae) Interaction with Geminivirus-Infected Host Plants: Bemisia tabaci, Host Plants and Geminiviruses (2011)
Spirodiclofen and spiromesifen
Nauen R, et al.
Pesticide Outlook null
Jian-Long Bi et al.
Insect science, 23(1), 88-93 (2014-11-21)
The carmine spider mite, Tetranychus cinnabarinus (Boisduval) and the twospotted spider mite, Tetranychus urticae Koch, are serious pests of strawberries and many other horticultural crops. Control of these pests has been heavily dependent upon chemical acaricides. Objectives of this study
N Kissoum et al.
Comparative biochemistry and physiology. Toxicology & pharmacology : CBP, 228, 108666-108666 (2019-11-15)
Spiromesifen (Oberon® 240 SC), a pesticide widely used to control pests like mites and whiteflies, was investigated using Drosophila melanogaster Meigen, 1830 (Diptera, Drosophilidae) as a model organism. The compound was applied topically at two concentrations (LC10: 21.45 and LC25:
Baseline susceptibility of Bemisia tabaci B biotype (Hemiptera: Aleyrodidae) populations from California and Arizona to spiromesifen
Prabhaker N, et al.
Journal of Economic Entomology, 101, 174-181 (2008)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service