Merck
  • Role of ALKBH1 in the Core Transcriptional Network of Embryonic Stem Cells.
Role of ALKBH1 in the Core Transcriptional Network of Embryonic Stem Cells.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology (2016-01-15)
Rune Ougland, Ida Jonson, Marivi Nabong Moen, Gaute Nesse, Gry Asker, Arne Klungland, Elisabeth Larsen
ABSTRACT
ALKBH1, an AlkB homologue in the 2-oxoglutarate and Fe2+ dependent hydroxylase family, is a histone dioxygenase that removes methyl groups from histone H2A. Studies of transgenic mice lacking Alkbh1 reveal that most Alkbh1-/- embryos die during embryonic development. Embryonic stem cells (ESCs) derived from these mice have prolonged expression of pluripotency markers and delayed induction of genes involved in neural differentiation, indicating that ALKBH1 is involved in regulation of pluripotency and differentiation. The aim of this study was to further investigate the role ALKBH1 in early development. Double-filter methods for nitrocellulose-filter binding, dot blot, enzyme-linked immunosorbent assay (ELISA), immonocytochemistry, cell culture and differentiation of mouse ESCs, Co-IP and miRNA analysis. We found that SOX2 and NANOG bind the ALKBH1 promoter, and we identified protein-protein interactions between ALKBH1 and these core transcription factors of the pluripotency network. Furthermore, lack of ALKBH1 affected the expression of developmentally important miRNAs, which are involved in the regulation of NANOG, SOX2 and neural differentiation. Our results suggest that ALKBH1 interacts with the core transcriptional pluripotency network of ESCs and is involved in regulation of pluripotency and differentiation.
Sigma-Aldrich
Retinoic acid, ≥98% (HPLC), powder