Merck
  • Enhanced lignocellulosic biomass hydrolysis by oxidative lytic polysaccharide monooxygenases (LPMOs) GH61 from Gloeophyllum trabeum.
Enhanced lignocellulosic biomass hydrolysis by oxidative lytic polysaccharide monooxygenases (LPMOs) GH61 from Gloeophyllum trabeum.
Enzyme and microbial technology (2015-07-04)
Sera Jung, Younho Song, Ho Myeong Kim, Hyeun-Jong Bae
ABSTRACT
Lignocellulose is a renewable resource that is extremely abundant, and the complete enzymatic hydrolysis of lignocellulose requires a cocktail containing a variety of enzyme groups that act synergistically. The hydrolysis efficiency can be improved by introducing glycoside hydrolase 61 (GH61), a new enzyme that belongs to the auxiliary activity family 9 (AA9). GH61was isolated from Gloeophyllum trabeum and cleaves the glycosidic bonds on the cellulose surface via oxidation of various carbons. In this study, we investigated the properties of GH61. GtGH61 alone did not exhibit any notable activity, but the synergistic activity of GtGH61 with xylanase (GtXyl10G) or cellulase (GtCel5B) showed efficient bioconversion rates of 56 and 174% in pretreated kenaf (Hibiscus cannabinus L.) and oak (Quercus spp.), respectively. Furthermore, the GtGH61 activity was strongly accelerated in the presence of cobalt Co(2+). Enzyme cocktails (GtXyl10G, GtCel5B, and GtGH61) increased the amount of sugar released by 7 and 6% for pretreated oak and kenaf, respectively, and the addition of Co(2+) stimulated bioconversion by 12 and 11% in pretreated oak and kenaf, respectively.
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, Grade I, ≥99% (GC), suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)