Merck
  • A peroxidase biomimetic system based on Fe3O4 nanoparticles in non-enzymatic sensors.
A peroxidase biomimetic system based on Fe3O4 nanoparticles in non-enzymatic sensors.
Talanta (2015-05-13)
Juan A Ramos Guivar, Edson G R Fernandes, Valtencir Zucolotto
ABSTRACT
Magnetic nanoparticles have been applied in many areas of nanomedicine. Sensing platforms based on this type of nanoparticles have received attention due to the relative low cost and biocompatibility. Biosensor is the most widely investigated type of analytical device. This type of sensor combines the physicochemical transduction with the incorporation of biological sensing components. Among the biological components, enzymes are the most commonly used as sensitive elements. However, natural enzymes may exhibit serious disadvantages such as lack of stability and loss of catalytic activity after immobilization. The study of enzymatic biomimetic systems are of great interest. This study reports the development of a new sensor composed of Fe3O4@CTAB and poly(sodium 4-styrenesulfonate) (PSS) films assembled via Layer-by-Layer (LbL) technique and used as peroxidase mimetic systems. Magnetic nanoparticles (MNps) were synthesized using thermal decomposition method and further dispersed to aqueous medium by ligand modification reaction using cetyltrimethylammonium bromide (CTAB). The amperometric detection limit of H2O2 was found to be ca. 103 µmol L(-1). By chronoamperometry, the peroxidase biomimetic sensor exhibited a linear response for H2O2 in the range from 100 µmol L(-1) to 1.8 mmol L(-1) (R(2) = 0.994) with sensitivity of 16 nA mol(-1) L. The apparent Michaelis-Menten constant was 5.3 mmol L(-1), comparable with some biosensors based on peroxidase enzyme. Moreover, the sensor presented a reproducibility of ca. 7.7% (n = 4) and their response (response time: 90 s) is not significantly affected in the presence of some interferents including K(+), Na(+), Cl(-), Mg(2+), Ca(2+), and Uric Acid.
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Potassium phosphate dibasic, anhydrous, for luminescence, for molecular biology, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Oleic acid, natural, FCC
Sigma-Aldrich
Oleic acid, technical grade, 90%
Sigma-Aldrich
Oleylamine, technical grade, 70%
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Sigma-Aldrich
Potassium phosphate dibasic, 99.95% trace metals basis
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
Potassium phosphate dibasic solution, 1.0 M
Sigma-Aldrich
Uric acid, ≥99%, crystalline
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Sigma-Aldrich
Uric acid, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
Potassium phosphate dibasic, reagent grade, ≥98.0%
Sigma-Aldrich
Potassium phosphate dibasic, meets USP testing specifications
Sigma-Aldrich
Oleic acid, BioReagent, suitable for cell culture
Sigma-Aldrich
Potassium hydroxide, anhydrous, ≥99.95% trace metals basis
SAFC
Oleylamine, ≥98% (primary amine)
Sigma-Aldrich
Cyclohexane, anhydrous, 99.5%
Sigma-Aldrich
Iron(III) acetylacetonate, 97%
Sigma-Aldrich
Iron(III) acetylacetonate, ≥99.9% trace metals basis