Merck
  • Identification of the Same Na(+)-Specific DNAzyme Motif from Two In Vitro Selections Under Different Conditions.
Identification of the Same Na(+)-Specific DNAzyme Motif from Two In Vitro Selections Under Different Conditions.
Journal of molecular evolution (2015-11-19)
Seyed-Fakhreddin Torabi, Yi Lu
ABSTRACT
We report an investigation of the functional relationship between two independently selected RNA-cleaving DNAzymes, NaA43, and Ce13, through in vitro selection. The NaA43 DNAzyme was obtained through a combination of gel-based and column-based in vitro selection in the presence of Na(+) and reported to be highly selective for Na(+) over other metal ions. The Ce13 DNAzyme was isolated via a gel-based method in the presence of Ce(4+) and found to be active with trivalent lanthanides, Y(3+) and Pb(2+). Despite completely different activities reported for the two DNAzymes, they share a high level of sequence similarity (~60% sequence identity). In this work, we systematically analyzed the activity of both DNAzymes to elucidate their potential functional relationship. We found that Na(+) is an obligate cofactor of the Ce13 DNAzyme and lanthanides cannot initiate the cleavage reaction in the absence of Na(+). Hence, we conclude that the Ce13 DNAzyme is a variant of the NaA43 DNAzyme that catalyzes reaction in the presence Na(+) and also utilizes lanthanides in a potentially allosteric manner. These results have identified a new DNAzyme motif that is not only remarkably Na(+)-specific, but also allows for design of novel allosteric DNAzymes for different biotechnological applications.
Product Description
Supelco
Potassium hydroxide solution, volumetric, 8.0 M KOH (8.0N)
Sigma-Aldrich
Potassium hydroxide, ≥85% KOH basis, pellets, white
Sigma-Aldrich
Potassium hydroxide, BioXtra, ≥85% KOH basis
Sigma-Aldrich
Potassium hydroxide, anhydrous, ≥99.95% trace metals basis
Sigma-Aldrich
MES solution, BioUltra, for molecular biology, 0.5 M in H2O
Sigma-Aldrich
Boric acid, 99.999% trace metals basis
Sigma-Aldrich
Boric acid, 99.97% trace metals basis
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Boric acid, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, BioUltra, for molecular biology, 99% (T)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
MES, low moisture content, ≥99% (titration)
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Boric acid, BioXtra, ≥99.5%
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Boric acid, suitable for electrophoresis, ≥99.5%
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Boric acid, tablet, 1 g boric acid per tablet
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Boric acid, BioReagent, for molecular biology, suitable for cell culture, suitable for plant cell culture, ≥99.5%
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Acrylamide solution, 40%, suitable for electrophoresis, sterile-filtered