Skip to Content
Merck
All Photos(1)

Key Documents

P4494

Supelco

Potassium hydroxide solution

8.0 M KOH (8.0N), volumetric solution suitable for titration

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
$114.00

$114.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
50 ML
$114.00

About This Item

Linear Formula:
KOH
CAS Number:
Molecular Weight:
56.11
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.21

$114.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Product Name

Potassium hydroxide solution, volumetric, 8.0 M KOH (8.0N)

quality

volumetric

concentration

8.0 M KOH (8.0N)

SMILES string

[K+].[O-][H]

InChI

1S/K.H2O/h;1H2/q+1;/p-1

InChI key

KWYUFKZDYYNOTN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application


  • Impact of Modifications from Potassium Hydroxide on Porous Semi-IPN Hydrogel Properties and Its Application in Cultivation.: This study explores how potassium hydroxide modifications enhance the properties of porous semi-interpenetrating polymer network (IPN) hydrogels, specifically for use in cultivation applications. These modifications improve the hydrogel′s mechanical strength and water retention capacity, making it a viable option for agricultural purposes. (Tuan et al., 2024).

  • Characterization of Bioadsorbents from Organic Municipal Waste.: The research investigates the preparation and characterization of bioadsorbents derived from organic municipal waste treated with potassium hydroxide. These bioadsorbents show enhanced adsorption capacities for pollutants, demonstrating potential for environmental cleanup applications. (Sołtysik et al., 2024).

  • Robust anion exchange membranes based on ionic liquid grafted chitosan/polyvinyl alcohol/quaternary ammonium functionalized silica for polymer electrolyte membrane fuel cells.: This article discusses the development of durable anion exchange membranes using potassium hydroxide-treated materials. The enhanced stability and conductivity of these membranes are crucial for improving the performance of polymer electrolyte membrane fuel cells. (Jang et al., 2024).

  • [Optimization and application of determination method of cholesterol in various dishes].: This study, written in Chinese, focuses on optimizing a method for determining cholesterol levels in different dishes using potassium hydroxide solution, which improves the accuracy and efficiency of the analytical process. (Chen et al., 2023).

  • Highly efficient sulfur-doped Ni(3)Fe electrocatalysts for overall water splitting: Rapid synthesis, mechanism and driven by sustainable energy.: The paper explores the use of potassium hydroxide in the rapid synthesis of sulfur-doped nickel-iron electrocatalysts. These electrocatalysts exhibit high efficiency in water splitting, driven by sustainable energy sources, offering significant advancements in renewable energy technologies. (Xu et al., 2024).


Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Khatereh Bahrpaima et al.
Biomolecules, 9(8) (2019-08-23)
In this work, 1-carboxypropyled (1-CPRLS) and 5-carboxypentyled lignosulfonates (5-CPELS) were synthesized using 2-chlorobutanoic acid and 6-chlorohexanoic acid as carboxylate group donors via SN1 and SN2 mechanisms, respectively. 1-Carboxypropyl and 5-carboxypentyl lignosulfonates with the charge densities of -3.45 and -2.94 meq
Andreu Fabregat et al.
Steroids, 78(3), 327-336 (2012-12-25)
The occurrence of several polyunsaturated testosterone related compounds (including 4,6-androstadien-3,17-dione and 4,6-androstadien-17β-ol-3-one) in urine after alkaline treatment of the sample has been recently reported. Although several experiments seem to indicate that they are testosterone metabolites, their origin is still unknown.
Hui Boon Teh et al.
Journal of chromatography. A, 1383, 112-120 (2015-02-05)
A new, highly sensitive and reliable two-dimensional matrix elimination ion chromatography (IC) method was developed for simultaneous detection of bromate, chlorite and five haloacetic acids. This method combined the conventional IC in first dimension with capillary IC in the second
Heng-guo Wang et al.
ChemSusChem, 6(1), 56-60 (2012-12-12)
Between the sheets: Sodium-ion batteries are an attractive, low-cost alternative to lithium-ion batteries. Nitrogen-doped porous carbon sheets are prepared by chemical activation of polypyrrole-functionalized graphene sheets. When using the sheets as anode material in sodium-ion batteries, their unique compositional and
K Y Foo et al.
Bioresource technology, 130, 696-702 (2013-01-22)
In this work, preparation of granular activated carbon from oil palm biodiesel solid residue, oil palm shell (PSAC) by microwave assisted KOH activation has been attempted. The physical and chemical properties of PSAC were characterized using scanning electron microscopy, volumetric

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service