Merck
  • Effects of grind size when alkaline treating corn residue and impact of ratio of alkaline-treated residue and distillers grains on performance of finishing cattle.
Effects of grind size when alkaline treating corn residue and impact of ratio of alkaline-treated residue and distillers grains on performance of finishing cattle.
Journal of animal science (2015-10-07)
A L Shreck, B L Nuttelman, C J Schneider, D B Burken, J L Harding, G E Erickson, T J Klopfenstein, M J Cecava
ABSTRACT
Two studies were conducted to optimize use of alkaline-treated corn stover and wheat straw and distillers grains as partial corn replacements. In Exp. 1, a finishing experiment used 30 pens (12 steers/pen) of calf-fed steers (initial BW = 374 ± 23.9 kg) with a 2 × 2 + 1 factorial arrangement of treatments with 6 replications per treatment. Factors were grind size, where corn stover was processed through a 2.54- or 7.62-cm screen, and chemical treatment (corn stover either fed in native, non-treated form [NT; 93.4% DM] or alkaline treated [AT; 5% CaO hydrated to 50% DM]). No interactions (P ≥ 0.38) were noted between grind size and chemical treatment. Feeding AT compared with NT improved (P ≤ 0.02) final BW, ADG, and G:F. Reducing grind size improved (P ≤ 0.01) ADG and G:F, and no interaction with chemical treatment was observed. Steers fed AT had similar DMI, ADG, G:F, and carcass characteristics compared with a 5% roughage control that contained 15 percentage units (DM basis) more corn. In Exp. 2, 60 individually fed steers (initial BW = 402 ± 61.4 kg) were randomly assigned to 10 diets. Six treatments evaluated 10, 25, or 40% dry-rolled corn (DRC), which was replaced with either a 2:1 or 3:1 ratio (DM basis) of modified distillers grains plus solubles (MDGS) and treated corn stover analyzed as a 2 × 3 factorial. An additional 3 treatments were added where a 3:1 ratio of MDGS:straw were compared with a 3:1 ratio of MDGS:stover. As DRC increased, G:F (P = 0.06) quadratically increased for 3:1 MDGS:stover diets. Increasing DRC increased (P = 0.07) G:F in treated stover diets, regardless of ratio. Increasing DRC increased (P = 0.10) ADG for 3:1 ratios for both straw and stover. Reducing grind size, feeding a maximum of 20% treated crop residue, and maintaining at least 25% corn in the diet are strategies for optimizing cattle performance when replacing dry-rolled and high-moisture corn with treated crop residues and distillers grains.
Sigma-Aldrich
Calcium oxide, 99.995% trace metals basis
Sigma-Aldrich
Estradiol, meets USP testing specifications
Sigma-Aldrich
Calcium oxide, nanopowder, <160 nm particle size (BET), 98%
Sigma-Aldrich
Calcium oxide, reagent grade
Sigma-Aldrich
Calcium oxide, ReagentPlus®, 99.9% trace metals basis
Sigma-Aldrich
Calcium oxide, anhydrous, powder, ≥99.99% trace metals basis