Skip to Content
Merck
All Photos(2)

Key Documents

451711

Sigma-Aldrich

Calcium oxide

anhydrous, powder, ≥99.99% trace metals basis

Synonym(s):

Lime, Quicklime

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$606.00

$606.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$606.00

About This Item

Linear Formula:
CaO
CAS Number:
Molecular Weight:
56.08
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

$606.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

anhydrous

Quality Level

Assay

≥99.99% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: calcium

bp

2850 °C (lit.)

density

3.3 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

O=[Ca]

InChI

1S/Ca.O

InChI key

ODINCKMPIJJUCX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

María Nazaret González-Alcaraz et al.
Journal of environmental management, 116, 72-80 (2013-01-08)
The aim of this study was to identify the effectiveness of liming in combination with vegetation for the recovery of slightly acidic, saline soils of eutrophic wetlands affected by mine wastes, under fluctuating flooding conditions. Simulated soil profiles were constructed
Sebastião Roberto Soares et al.
Waste management (New York, N.Y.), 33(1), 175-183 (2012-11-06)
The establishment of rules to manage Health Care Waste (HCW) is a challenge for the public sector. Regulatory agencies must ensure the safety of waste management alternatives for two very different profiles of generators: (1) hospitals, which concentrate the production
Vasilije Manovic et al.
International journal of environmental research and public health, 7(8), 3129-3140 (2010-10-16)
This paper presents a review of the research on CO(2) capture by lime-based looping cycles undertaken at CanmetENERGY's (Ottawa, Canada) research laboratories. This is a new and very promising technology that may help in mitigation of global warming and climate
Kunlun Zhang et al.
Journal of hazardous materials, 243, 278-285 (2012-11-20)
Tetrabromobisphenol A (TBBPA) is the most widely used brominated flame retardant (BFR), which has received more and more concerns due to its high lipophilicity, persistency and endocrine disrupting property in the environment. Considering the possible need for the safe disposal
Arup Roy et al.
Journal of biomedical nanotechnology, 9(9), 1570-1578 (2013-08-29)
The high degree of microbial diseases and their multidrug resistant properties make the researchers to develop new class of antimicrobial agents. A modern and innovative approach of drug development is the use of metallic nanoparticles as new formulations of antimicrobial

Articles

Three approaches generate white light, including LED-based down-conversion for broader applications.

Questions

  1. In #451711, 1) Anhydrous CaO means there's "Zero moisture" Calcium Oxides ? 2) the purity 99.99% means there's 0.01% impurities ?

    1 answer
    1. Product 451711 does not have a specification for water content. This is likely because calcium oxide reacts with water to form calcium hydroxide, so water would not be detected. The term "anhydrous" is used more to distinguish this Calcium oxide product (typically approximately 71 percent Calcium by weight) from Calcium hydroxide (typically approximately 54 percent Calcium by weight).
      The purity value of 99.99 percent is on a trace metals basis. That is, Product 451711 contains AT MOST 0.01 percent or 100 ppm of metals other than Calcium.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service