Merck
  • Histone Deacetylase 10 Regulates the Cell Cycle G2/M Phase Transition via a Novel Let-7-HMGA2-Cyclin A2 Pathway.
Histone Deacetylase 10 Regulates the Cell Cycle G2/M Phase Transition via a Novel Let-7-HMGA2-Cyclin A2 Pathway.
Molecular and cellular biology (2015-08-05)
Yixuan Li, Lirong Peng, Edward Seto
ABSTRACT
Histone deacetylase (HDAC) inhibition leads to cell cycle arrest in G1 and G2, suggesting HDACs as therapeutic targets for cancer and diseases linked to abnormal cell growth and proliferation. Many HDACs are transcriptional repressors. Some may alter cell cycle progression by deacetylating histones and repressing transcription of key cell cycle regulatory genes. Here, we report that HDAC10 regulates the cell cycle via modulation of cyclin A2 expression, and cyclin A2 overexpression rescues HDAC10 knockdown-induced G2/M transition arrest. HDAC10 regulates cyclin A2 expression by deacetylating histones near the let-7 promoter, thereby repressing transcription. In HDAC10 knockdown cells, let-7f and microRNA 98 (miR-98) were upregulated and the let-7 family target, HMGA2, was downregulated. HMGA2 loss resulted in enrichment of the transcriptional repressor E4F at the cyclin A2 promoter. These findings support a role for HDACs in cell cycle regulation, reveal a novel mechanism of HDAC10 action, and extend the potential of HDACs as targets in diseases of cell cycle dysregulation.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Hdac6
Sigma-Aldrich
MISSION® esiRNA, targeting human HDAC6
Sigma-Aldrich
Thymidine, ≥99.0% (HPLC)
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Thymidine, ≥99%
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ≥95% (HPLC)
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ~98% (HPLC)
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., suitable for cell culture, ≥95%
Sigma-Aldrich
RO-3306, ≥98% (HPLC)
Sigma-Aldrich
Thymidine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Anti-GAPDH antibody, Mouse monoclonal, clone GAPDH-71.1, purified from hybridoma cell culture
Sigma-Aldrich
Hydroxyurea, 98%, powder
Sigma-Aldrich
Trichostatin A, ≥98% (HPLC), from Streptomyces sp.
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2
Sigma-Aldrich
DAPI, for nucleic acid staining