Merck
  • Expansion of the HSFY gene family in pig lineages : HSFY expansion in suids.
Expansion of the HSFY gene family in pig lineages : HSFY expansion in suids.
BMC genomics (2015-06-10)
Benjamin M Skinner, Kim Lachani, Carole A Sargent, Fengtang Yang, Peter Ellis, Toby Hunt, Beiyuan Fu, Sandra Louzada, Carol Churcher, Chris Tyler-Smith, Nabeel A Affara
ABSTRACT
Amplified gene families on sex chromosomes can harbour genes with important biological functions, especially relating to fertility. The Y-linked heat shock transcription factor (HSFY) family has become amplified on the Y chromosome of the domestic pig (Sus scrofa), in an apparently independent event to an HSFY expansion on the Y chromosome of cattle (Bos taurus). Although the biological functions of HSFY genes are poorly understood, they appear to be involved in gametogenesis in a number of mammalian species, and, in cattle, HSFY gene copy number may correlate with levels of fertility. We have investigated the HSFY family in domestic pig, and other suid species including warthog, bushpig, babirusa and peccaries. The domestic pig contains at least two amplified variants of HSFY, distinguished predominantly by presence or absence of a SINE within the intron. Both these variants are expressed in testis, and both are present in approximately 50 copies each in a single cluster on the short arm of the Y. The longer form has multiple nonsense mutations rendering it likely non-functional, but many of the shorter forms still have coding potential. Other suid species also have these two variants of HSFY, and estimates of copy number suggest the HSFY family may have amplified independently twice during suid evolution. The HSFY genes have become amplified in multiple species lineages independently. HSFY is predominantly expressed in testis in domestic pig, a pattern conserved with cattle, in which HSFY may play a role in fertility. Further investigation of the potential associations of HSFY with fertility and testis development may be of agricultural interest.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
2,4-Dinitrophenol, moistened with water, ≥98.0%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture