Merck
  • Comparison of proteome typing and serotyping of Streptococcus parauberis isolates from olive flounder (Paralichthys olivaceus).
Comparison of proteome typing and serotyping of Streptococcus parauberis isolates from olive flounder (Paralichthys olivaceus).
Journal of microbiological methods (2015-10-02)
Si Won Kim, Ho Bin Jang, Jung Seok Lee, Se Pyeong Im, Jassy Mary S Lazarte, Jong Pyo Seo, Woo Jai Lee, Jae Sung Kim, Tae Sung Jung
ABSTRACT
The olive flounder (Paralichthys olivaceus) is a cultivated marine species that is economically important in Korea and Japan. Several bacterial pathogens have caused severe mortalities in farmed olive flounder, especially Streptococcus parauberis. We collected 145 S. parauberis isolates from diseased olive flounders from 2003 to 2008 in Jeju Island, South Korea and characterized them by Matrix-assisted laser desorption ionization-time of flight mass spectrometry (MALDI TOF MS) and by serology. The serological analysis divided the isolates into serotype I (62.1%) and serotype II (36.6%) and the proteome analysis divided the isolates into cluster 1 (43.4%) and cluster 2 (56.6%). All cluster 1 isolates had serotype I, but cluster 2 consisted of serotype I (32.9%), serotype II (64.6%), and others (2.5%). Further detailed analysis of the mass spectra led to identification of several specific m/z peaks that enabled discrimination between cluster 1 and 2 and between serotype I and II within cluster 2. Our results suggest that MALDI TOF MS analysis has potential as an alternative method for the rapid and reliable identification of the fish pathogen S. parauberis.
Product Description
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, suitable for MALDI-TOF MS
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, 99%
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, 97%
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, ≥98% (TLC), powder
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Trifluoroacetic acid, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Sigma-Aldrich
Acetonitrile, for preparative HPLC, ≥99.8% (GC)
Sigma-Aldrich
Trifluoroacetic acid, ≥99%, for protein sequencing
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%