Merck
  • A multi-targeted liquid chromatography-mass spectrometry screening procedure for the detection in human urine of drugs non-prohibited in sport commonly used by the athletes.
A multi-targeted liquid chromatography-mass spectrometry screening procedure for the detection in human urine of drugs non-prohibited in sport commonly used by the athletes.
Journal of pharmaceutical and biomedical analysis (2015-09-08)
Monica Mazzarino, Lorenzo Cesarei, Xavier de la Torre, Ilaria Fiacco, Paul Robach, Francesco Botrè
ABSTRACT
This work presents an analytical method for the simultaneous analysis in human urine of 38 pharmacologically active compounds (19 benzodiazepine-like substances, 7 selective serotonin reuptake inhibitors, 4 azole antifungal drugs, 5 inhibitors of the phosphodiesterases type 4 and 3 inhibitors of the phosphodiesterase type 5) by liquid-chromatography coupled with tandem mass spectrometry. The above substances classes include both the most common "non banned" drugs used by the athletes (based on the information reported on the "doping control form") and those drugs who are suspected to be performance enhancing and/or act as masking agents in particular conditions. The chromatographic separation was performed by a reverse-phase octadecyl column using as mobile phases acetonitrile and ultra-purified water, both with 0.1% formic acid. The detection was carried out using a triple quadrupole mass spectrometric analyser, positive electro-spray as ionization source and selected reaction monitoring as acquisition mode. Sample pre-treatment consisted in an enzymatic hydrolysis followed by a liquid-liquid extraction in neutral field using tert-butyl methyl-ether. The analytical procedure, once developed, was validated in terms of sensitivity (lower limits of detection in the range of 1-50 ng mL(-1)), specificity (no interferences were detected at the retention time of all the analytes under investigation), recovery (≥60% with a satisfactory repeatability, CV % lower than 10), matrix effect (lower than 30%) and reproducibility of retention times (CV% lower than 0.1) and of relative abundances (CV% lower than 15). The performance and the applicability of the method was evaluated by analyzing real samples containing benzodiazepines (alprazolam, diazepam, zolpidem or zoplicone) or inhibitors of the phosphodiesterases type 5 (sildenafil or vardenafil) and samples obtained incubating two of the phosphodiesterases type 4 studied (cilomilast or roflumilast) with pooled human liver microsomes. All the parent compounds, together with their main phase I metabolites, were clearly detected using the analytical procedures here developed.
Sigma-Aldrich
tert-Butyl methyl ether, reagent grade, 98%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
tert-Butyl methyl ether, ACS reagent, ≥99.0%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
tert-Butyl methyl ether, reagent grade, ≥98%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
tert-Butyl methyl ether, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
tert-Butyl methyl ether, anhydrous, 99.8%
Sigma-Aldrich
Pyrrole, reagent grade, 98%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
Sigma-Aldrich
Fluconazole, ≥98% (HPLC), powder
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Pyrrole, ≥98%, FCC, FG
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Sodium phosphate dibasic, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E 339, anhydrous, 98-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%, free-flowing, Redi-Dri
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Magnesium chloride hexahydrate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Magnesium chloride hexahydrate, ACS reagent, 99.0-102.0%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Sodium phosphate dibasic, ACS reagent, ≥99.0%