Skip to Content
Merck
All Photos(1)

Key Documents

04276

Sigma-Aldrich

Sodium phosphate dibasic

puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E 339, anhydrous, 98-100.5% (calc. to the dried substance)

Synonym(s):

sec-Sodium phosphate, Disodium hydrogen phosphate, Disodium phosphate, Sodium hydrogenphosphate

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
$268.00
5 KG
$562.00
25 KG
inquire

$268.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 KG
$268.00
5 KG
$562.00
25 KG
inquire

About This Item

Linear Formula:
Na2HPO4
CAS Number:
Molecular Weight:
141.96
EC Number:
MDL number:
UNSPSC Code:
12161700
eCl@ss:
38070211
PubChem Substance ID:
NACRES:
NA.25
form:
powder
solubility:
H2O: soluble 8 parts
H2O: very soluble (Hot)
alcohol: insoluble

$268.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Agency

meets analytical specifications of BP
meets analytical specifications of Ph. Eur.
meets analytical specifications of USP

grade

puriss.

Assay

98-100.5% (calc. to the dried substance)

form

powder

quality

meets analytical specification of Ph. Eur., BP, USP, FCC, E 339

impurities

reducing matter, complies
residual solvents, complies
≤0.001% heavy metals (as Pb)
≤0.002% ammonium (NH4)
≤0.2% water-insoluble matter
≤1% NaH2PO4 (calc. to dried subst.)
≤2.0% pyrophosphate (as Na4P2O7)

loss

≤1% loss on drying, 105°C, 4h
≤1% loss on drying, 130 °C

pH

8.4-9.6 (20 °C, 1%)

pKa (25 °C)

(1) 2.15, (2) 6.82, (3) 12.38 (phosphoric acid)

solubility

H2O: soluble 8 parts
H2O: very soluble (Hot)
alcohol: insoluble

anion traces

chloride (Cl-): ≤50 mg/kg
fluoride (F-): ≤10 mg/kg
sulfate (SO42-): ≤200 mg/kg

cation traces

As: ≤2 mg/kg
Cd: ≤1 mg/kg
Cu: ≤10 mg/kg
Fe: ≤20 mg/kg
Hg: ≤1 mg/kg
Pb: ≤4 mg/kg
Zn: ≤10 mg/kg

suitability

complies for appearance of solution

SMILES string

[Na+].[Na+].[H]OP([O-])([O-])=O
[Na+].[Na+].[H]OP([O-])([O-])=O

InChI

1S/2Na.H3O4P/c;;1-5(2,3)4/h;;(H3,1,2,3,4)/q2*+1;/p-2

InChI key

BNIILDVGGAEEIG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Sodium phosphate dibasic has been used for preparing phosphate buffer which in turn is utilized in X-gal staining.[1]

Biochem/physiol Actions

Sodium phosphate dibasic is an important component of running buffer of denaturing gel electrophoresis.[2] It induces glomerular calcification as well as other nephrotic syndrome in Sprague-Dawley rats.[3]

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Duvillié et al.
Endocrinology, 143(4), 1530-1537 (2002-03-19)
The targeted disruption of the two nonallelic insulin genes in mouse was reported previously to result in intrauterine growth retardation, severe diabetes immediately after suckling, and death within 48 h of birth. We have further used these animals to investigate
Noriko Tsuchiya et al.
Toxicologic pathology, 32(4), 408-412 (2004-06-19)
To elucidate the nephrotoxicity of phosphate, dibasic sodium phosphate solution was given to Sprague-Dawley rats by daily bolus intravenous administration at concentrations of 0, 1, 25, 250, or 360 mM (0, 1, 28, 284, or 408 mg/kg Na2HPO4) for 14
Noriko Tsuchiya et al.
Toxicologic pathology, 37(3), 270-279 (2009-02-27)
Sprague-Dawley rats received once daily tail-vein injections of 360 mM dibasic sodium phosphate solution at 8 mL/kg for fourteen or twenty-eight days. Clinical examination revealed persistent proteinuria from three days after the first dosing and thereafter severe proteinuria from eight
Megan A Rees et al.
Journal of proteome research, 14(1), 120-132 (2014-10-21)
Interactions between a host and a bacterial pathogen are mediated by cross-talk between molecules present on, or secreted by, pathogens and host binding-molecules. Identifying proteins involved at this interface would provide substantial insights into this interaction. Although numerous studies have
Samuel Adler et al.
Gastrointestinal endoscopy, 76(6), 1170-1174 (2012-10-03)
Colon capsule endoscopy (CCE) is a noninvasive technique for the detection of colorectal lesions. However, for CCE to be offered as an out-of-clinic procedure, the system needs to automatically alert the patient when to ingest the laxative (booster). We tested

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service