Merck
  • Enzymatic Analysis of Positional Fatty Acid Distributions in Triacylglycerols by 1(3)-Selective Transesterification with Candida antarctica Lipase B: a Collaborative Study.
Enzymatic Analysis of Positional Fatty Acid Distributions in Triacylglycerols by 1(3)-Selective Transesterification with Candida antarctica Lipase B: a Collaborative Study.
Journal of oleo science (2015-11-03)
Yomi Watanabe, Shinichi Sato, Mihoko Asada, Toshiharu Arishima, Yasuhiro Iida, Jun Imagi, Katsuyoshi Saito, Takashi Sano, Akiko Sasaki, Ryo Sasaki, Chiemi Sato, Tadahisa Shibuya, Yuki Tsukahara, Toshiharu Nagai, Toru Fukazawa, Ryuji Hori, Rika Homma, Yosuke Miyazaki, Atsushi Yamashita, Kazuaki Yoshinaga, Shimpei Watanabe
ABSTRACT
The positional distributions of fatty acids (FAs) in fats and oils are principally analyzed by selectively transesterifying the target triacylglycerols (TAGs) at the 1(3) position using Pseudozyma (Candida) antarctica lipase, followed by recovering the resulting 2-monoacylglycerols (MAGs) by chromatography. FA compositions were measured by gas chromatography (GC) after methylating target TAGs and 2-MAGs. The method was collaboratively evaluated by 12 laboratories by analyzing the positional FA distributions in soybean, palm, and sardine oils. The maximum reproducibility relative standard deviations for the major FAs and those at the sn-2 positions of soybean, palm, and sardine oils were 4.41% and 3.92% (18:3n-3), 4.48% and 3.82% (18:0), and 8.93 and 8.24% (14:0), respectively. The values at the sn-2 position were always low. Therefore, these results indicated that the variations were mainly caused by the FA analysis procedure, i.e., the methylation and GC analyses, rather than the enzymatic transesterification and chromatography utilized to prepare 2-MAGs from the target oil.
Product Description
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Sodium methoxide, reagent grade, 95%, powder
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Sodium methoxide solution, 25 wt. % in methanol
Sigma-Aldrich
Sodium methoxide solution, ACS reagent, 0.5 M CH3ONa in methanol (0.5N)
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Potassium hydroxide, free-flowing, Redi-Dri, ACS reagent, ≥85%, pellets
Sigma-Aldrich
Potassium hydroxide, anhydrous, ≥99.95% trace metals basis
Sigma-Aldrich
Potassium hydroxide, ACS reagent, ≥85%, pellets
Supelco
Potassium hydroxide solution, volumetric, 8.0 M KOH (8.0N)
Sigma-Aldrich
Potassium hydroxide, ≥85% KOH basis, pellets, white
Sigma-Aldrich
Potassium hydroxide, BioXtra, ≥85% KOH basis
Sigma-Aldrich
Potassium hydroxide solution, 45 wt. % in H2O
Sigma-Aldrich
Potassium hydroxide, puriss. p.a., ≥86% (T), pellets
Sigma-Aldrich
Potassium hydroxide, reagent grade, 90%, flakes
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, for molecular biology
Sigma-Aldrich
Potassium hydroxide, semiconductor grade, pellets, 99.99% trace metals basis (Purity excludes sodium content.)
Sigma-Aldrich
Potassium hydroxide, pellets, reag. Ph. Eur., ≥85%