Merck
  • A high throughput screening system for predicting chemically-induced reproductive organ deformities.
A high throughput screening system for predicting chemically-induced reproductive organ deformities.
Reproductive toxicology (Elmsford, N.Y.) (2014-12-22)
Bart van der Burg, Bart Pieterse, Harrie Buist, Geertje Lewin, Sander C van der Linden, Hai-yen Man, Emiel Rorije, Aldert H Piersma, Inge Mangelsdorf, Andre P M Wolterbeek, E Dinant Kroese, Barbara van Vugt-Lussenburg
ABSTRACT
There is a great need for alternative testing methods for reproductive toxicants that are practical, fast, cost-effective and easy to interpret. Previously we followed a pragmatic approach using readily available tests, which was successful in predicting reproductive toxicity of chemicals [13]. This initial battery still contained apical tests and is fairly complex and low in its throughput. The current study aimed to simplify this screening battery using a mechanistic approach and a panel of high throughput CALUX reporter gene assays. A mechanistic approach was taken to validate this high throughput test battery. To this end it was challenged with two preselected sets of chemicals addressing two major apical effect classes relevant in reproductive toxicity. We found selectivity in this battery in that 82% of the compounds inducing reproductive organ deformities were predicted correctly, while for compounds inducing neural tube defects this was the case in 47% only. This is consistent with the mechanisms of toxicity covered in the battery. The most informative assays in the battery were ERalpha CALUX to measure estrogenicity and the AR-anti CALUX assay to measure androgen receptor antagonism.
Sigma-Aldrich
Dibutyltin dichloride, 96%
Sigma-Aldrich
Diisobutyl phthalate, 99%
Sigma-Aldrich
Diethylene glycol, ReagentPlus®, 99%
Sigma-Aldrich
Benzyl butyl phthalate, 98%
Sigma-Aldrich
Glycolic acid, ReagentPlus®, 99%
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
Diethylene glycol, BioUltra, ≥99.0% (GC)
Sigma-Aldrich
Diethylene glycol, puriss. p.a., ≥99.0% (GC), colorless
Sigma-Aldrich
Methyl 1-(butylcarbamoyl)-2-benzimidazolecarbamate, 95%
Sigma-Aldrich
Glycolic acid solution, technical grade, 70 wt. % in H2O
Sigma-Aldrich
Diethylene glycol dimethyl ether, anhydrous, 99.5%
Sigma-Aldrich
Glycolic acid solution, high purity, 70 wt. % in H2O
Sigma-Aldrich
Acetylsalicylic acid, analytical standard
Sigma-Aldrich
Glycolic acid, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
Diethylstilbestrol, ≥99% (HPLC)
Sigma-Aldrich
Acetylsalicylic acid, ≥99.0%
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
2-Methoxyethanol, anhydrous, 99.8%
Sigma-Aldrich
Cadmium chloride, 99.99% trace metals basis
Sigma-Aldrich
2-Ethylhexanoic acid, ≥99%
Sigma-Aldrich
Nonylphenol, technical grade, mixture of ring and chain isomers
Sigma-Aldrich
Tributyltin chloride, 96%
Sigma-Aldrich
Phenol Red, ACS reagent
Sigma-Aldrich
2-Ethylhexanoic acid, 99%
Sigma-Aldrich
Bisphenol A, 97%
Supelco
Bisphenol A, ≥99%