Merck
  • A colorimetric probe to determine Pb(2+) using functionalized silver nanoparticles.
A colorimetric probe to determine Pb(2+) using functionalized silver nanoparticles.
The Analyst (2015-11-12)
Kwon-Chul Noh, Yun-Sik Nam, Ho-Jin Lee, Kang-Bong Lee
ABSTRACT
A simple and sensitive colorimetric method for the determination of Pb(2+) ions in aqueous samples was developed using 1-(2-mercaptoethyl)-1,3,5-triazinane-2,4,6-trione (MTT) functionalized silver nanoparticles (MTT-AgNPs). The Pb(2+) ion acted as the metal center of the coordination complex, which formed N-Pb(2+)-O coordination bonds with the MTT-AgNPs, shortening the interparticle distance, and inducing aggregation of the MTT-AgNPs. This aggregation resulted in a dramatic color change from yellow to dark blue. Using this methodology, the concentration of Pb(2+) ions in environmental samples could be quantitatively detected with the naked eye or by using UV-vis spectrometry. Also, we found that the selectivity and sensitivity of detection were noticeably improved in the pH range of 7-8, at which a more obvious color change was observed. The absorption ratios (A625/A395) of the modified AgNP solution exhibited a linear correlation with Pb(2+) ion concentrations within the linear range of 0.1-0.6 μg mL(-1), and the limits of detection in tap and pond water were 0.02 and 0.06 μg mL(-1), respectively. This cost-effective sensing system allows for the rapid and facile determination of Pb(2+) ions in aqueous samples.
Sigma-Aldrich
VenPure® SF, powder
Sigma-Aldrich
Sodium borohydride solution, 2.0 M in triethylene glycol dimethyl ether
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Sodium borohydride solution, ~12 wt. % in 14 M NaOH
Sigma-Aldrich
Potassium thiocyanate, BioXtra, ≥99.0%
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Potassium thiocyanate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Potassium thiocyanate, ACS reagent, 99%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium borohydride, purum p.a., ≥96% (gas-volumetric)
Sigma-Aldrich
Sodium borohydride, ReagentPlus®, 99%
Sigma-Aldrich
Sodium borohydride, caplets (18 × 10 × 8 mm), 98%
Sigma-Aldrich
Sodium borohydride, granular, 10-40 mesh, 98%
Sigma-Aldrich
Sodium borohydride, granular, 99.99% trace metals basis
Sigma-Aldrich
Hydriodic acid, 57 wt. %, distilled, 99.999% trace metals basis
Sigma-Aldrich
Sodium borohydride, powder, ≥98.0%
Sigma-Aldrich
Hydriodic acid, contains ≤1.5% hypophosphorous acid as stabilizer, ACS reagent, ≥47.0%
Sigma-Aldrich
Hydriodic acid, contains no stabilizer, distilled, 57 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Hydriodic acid, 57 wt. % in H2O, distilled, stabilized, 99.95%
Sigma-Aldrich
Hydriodic acid, contains no stabilizer, ACS reagent, 55%