Skip to Content
Merck
All Photos(1)

Key Documents

P2713

Sigma-Aldrich

Potassium thiocyanate

BioXtra, ≥99.0%

Synonym(s):

Potassium rhodanide

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
$172.00
1 KG
$546.00

$172.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 G
$172.00
1 KG
$546.00

About This Item

Linear Formula:
KSCN
CAS Number:
Molecular Weight:
97.18
Beilstein:
3594799
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.31
grade:
for analytical purposes
form:
crystals
solubility:
H2O: 1 M, clear, colorless

$172.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

for analytical purposes

Quality Level

product line

BioXtra

Assay

≥99.0%

form

crystals

impurities

≤0.001% Phosphorus (P)
≤0.005% Insoluble matter

mp

173 °C (lit.)

solubility

H2O: 1 M, clear, colorless

anion traces

chloride (Cl-): ≤0.005%
sulfate (SO42-): ≤0.005%

cation traces

Al: ≤0.005%
Ca: ≤0.001%
Cu: ≤0.0005%
Fe: ≤0.0005%
Mg: ≤0.0005%
NH4+: ≤0.003%
Na: ≤0.005%
Pb: ≤0.001%
Zn: ≤0.0005%

SMILES string

[K]SC#N

InChI

1S/CHNS.K/c2-1-3;/h3H;/q;+1/p-1

InChI key

ZNNZYHKDIALBAK-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Potassium thiocyanate has been used as one of the component in the formulation of artificial saliva for in vitro human digestion model.[1][2] It has also been used in the isolation of glycosaminoglycans from fish.[3]

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1

Supplementary Hazards

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dietary glycosaminoglycans interfere in bacterial adhesion and gliadin-induced pro-inflammatory response in intestinal epithelial (Caco-2) cells
Laparra JM, et al.
International Journal of Biological Macromolecules, 47(4), 458-464 (2010)
Bioaccessibility of Hg, Cd and As in cooked black scabbard fish and edible crab
Maulvault A, et al.
Food And Chemical Toxicology, 49(11), 2808-2815 (2011)
German Cano-Sancho et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 86, 34-40 (2015-09-27)
This study was aimed at characterizing the bioaccessibility of arsenic (As), mercury (Hg) and methylmercury (MeHg) in commercial fish and seafood species widely consumed by the population of Catalonia (Spain). An accurate evaluation on how bioaccessibility estimations may affect the
A Botti et al.
The journal of physical chemistry. B, 113(29), 10014-10021 (2009-07-03)
The results of a neutron diffraction experiment performed on aqueous solutions of KSCN are analyzed and discussed in comparison with similar data for KCl and KOH solutions. The effect of the different ions on the structure of water and the
Hongtao Bian et al.
The journal of physical chemistry. B, 116(49), 14426-14432 (2012-11-28)
Microscopic structures and dynamics of aqueous salt solutions were investigated with the ultrafast vibrational energy exchange method and anisotropy measurements. In KSCN aqueous solutions of various concentrations, the rotational time constants of SCN(-) anions are proportional to the viscosities of

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service