Merck
  • Patch clamp characterization of the effect of cardiolipin on MscS of E. coli.
Patch clamp characterization of the effect of cardiolipin on MscS of E. coli.
European biophysics journal : EBJ (2015-04-07)
Pietro Ridone, Yoshitaka Nakayama, Boris Martinac, Andrew R Battle
ABSTRACT
The bacterial mechanosensitive channels MscS and MscL are gated by an increase in membrane tension when the bacterium experiences hypoosmotic shock. It has been well established that membrane lipids modulate the mechanosensitivity and gating behavior of these channels. The focus of this study is a negatively charged phospholipid, cardiolipin, which has been shown to localize at curved regions of the bacterial cell, including the poles and the septum, and to have a strong preference for binding to membrane proteins. Here we characterize the effect of cardiolipin on MscS, the mechanosensitive channel of small conductance, using patch-clamp electrophysiology. We compare the gating kinetics and mechanosensitivity of the channel in both azolectin and mixtures of pure lipids DOPE/DOPC liposomes with and without cardiolipin. In azolectin liposomes, the addition of 10 % cardiolipin abolishes hysteresis of MscS, but MscL remains largely unaffected, indicating that cardiolipin may stabilize the closed state of MscS. On the other hand, mixtures of DOPE/DOPC abolish the hysteresis gating of MscS even in the absence of cardiolipin, and the addition of cardiolipin increases the opening and closing thresholds of both MscS and MscL. In addition, we show that MscS gates more frequently when cardiolipin is present in both the azolectin and pure lipid systems; this dose-dependent effect ultimately destabilizes the open state of MscS and we consider the functional implications of this cardiolipin effect in the bacterial osmotic response. Our results show that cardiolipin modulates the mechanosensitivity and gating characteristics of MscS, indicating its important role in the physiology of bacterial cells.
Sigma-Aldrich
Imidazole, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Imidazole, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Imidazole, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
L-α-Phosphatidylcholine, from soybean, Type II-S, 14-29% choline basis
Sigma-Aldrich
n-Dodecyl β-D-maltoside, ≥98% (GC)
Sigma-Aldrich
Imidazole, ACS reagent, ≥99% (titration)
Sigma-Aldrich
Imidazole, ≥99% (titration), crystalline
Sigma-Aldrich
L-α-Phosphatidylethanolamine, dioleoyl, ≥99% (GC), ≥98% (TLC), lyophilized powder
Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration)
Sigma-Aldrich
Ampicillin, meets USP testing specifications
Sigma-Aldrich
n-Dodecyl β-D-maltoside, BioXtra, ≥98% (GC)
Sigma-Aldrich
Imidazole, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%, Redi-Dri, free-flowing
Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration), free-flowing, Redi-Dri
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Imidazole buffer Solution, BioUltra, 1 M in H2O
Sigma-Aldrich
Isopropyl β-D-1-thiogalactopyranoside, ≥99% (TLC)
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Chloramphenicol, meets USP testing specifications
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
IPTG, ≥99% (TLC), ≤0.1% Dioxane
Sigma-Aldrich
Chloramphenicol, BioReagent, suitable for plant cell culture
Sigma-Aldrich
Chloramphenicol, ≥98% (HPLC)