Merck
  • Immobilization of Escherichia coli cells expressing 4-oxalocrotonate tautomerase for improved biotransformation of β-nitrostyrene.
Immobilization of Escherichia coli cells expressing 4-oxalocrotonate tautomerase for improved biotransformation of β-nitrostyrene.
Bioprocess and biosystems engineering (2015-09-28)
Lidija Djokic, Jelena Spasic, Sanja Jeremic, Branka Vasiljevic, Olivera Prodanovic, Radivoje Prodanovic, Jasmina Nikodinovic-Runic
ABSTRACT
The enzyme 4-oxalocrotonate tautomerase (4-OT) encoded by the xylH gene is a part of the degradation pathway of aromatic compounds in Pseudomonas putida mt-2. 4-OT was described to catalyze Michael-type addition of acetaldehyde to β-nitrostyrene, and the whole cell system based on recombinantly expressed 4-OT has been developed previously. In this study biocatalytic process based on Escherichia coli whole cells expressing 4-OT was significantly improved using immobilization and ex situ product recovery strategies. Whole cell immobilization in alginate beads was applied in biocatalytic production of 4-nitro-3-phenyl-butanal from β-nitrostyrene and acetaldehyde. Immobilized biocatalyst showed wider pH activity range and could tolerate twofold higher initial concentrations of substrate in comparison to the free whole cell biocatalyst. Beads retained their initial activity over 10 consecutive biotransformations of the model reaction and remained suitable for the repetitive use with 85% of the initial activity after two months of storage. Bioprocess was further improved by utilizing Amberlite XAD-2 hydrophobic resin for the product recovery. With this modification, the amount of organic solvent was reduced 40-fold in comparison to previously reported method making this biocatalytic process greener.
Sigma-Aldrich
Acetaldehyde solution, 50 wt. % in ethanol
Sigma-Aldrich
Acetaldehyde solution, natural, 50 wt. % ethanol, FG
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Ampicillin, meets USP testing specifications
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Acetaldehyde solution, 40 wt. % in H2O
Sigma-Aldrich
Acetaldehyde solution, 5 M in THF
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl acetate, biotech. grade, ≥99.8%
Sigma-Aldrich
Ethyl acetate, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99.5% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, for molecular biology
Sigma-Aldrich
Ethyl acetate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Supelco
Acetaldehyde, PESTANAL®, analytical standard
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Acetaldehyde, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Sigma-Aldrich
IPTG, ≥99% (TLC), ≤0.1% Dioxane
Sigma-Aldrich
Isopropyl β-D-1-thiogalactopyranoside, ≥99% (TLC)
Sigma-Aldrich
Isopropyl β-D-thiogalactopyranoside solution, ReadyMade IPTG solution for Blue-white screening
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetaldehyde, natural, FG
SAFC
Isopropyl β-D-1-thiogalactopyranoside