Merck
  • The binding selectivity of vonoprazan (TAK-438) to the gastric H+, K+ -ATPase.
The binding selectivity of vonoprazan (TAK-438) to the gastric H+, K+ -ATPase.
Alimentary pharmacology & therapeutics (2015-10-02)
D R Scott, K B Munson, E A Marcus, N W G Lambrecht, G Sachs
ABSTRACT
The gastric H(+) ,K(+) -ATPase is the preferred target for acid suppression. Until recently, the only drugs that effectively inhibited this ATPase were the proton pump inhibitors (PPIs). PPIs are acid-activated prodrugs that require acid protection. Once acid-activated, PPIs bind to cysteines of the ATPase, resulting in covalent, long-lasting inhibition. The short plasma half-life of PPIs and continual de novo synthesis of the H(+) ,K(+) -ATPase result in difficulty controlling night-time acid secretion. A new alternative to PPIs is the pyrrolo-pyridine, vonoprazan (TAK-438), a potassium-competitive acid blocker (PCAB) that does not require acid protection. In contrast to other PCABs, vonoprazan has a long duration of action, resulting in 24-h control of acid secretion, a high pKa of 9.37 and high affinity (Ki = 3.0 ηmol/L). To determine binding selectivity of vonoprazan for the gastric H(+) ,K(+) -ATPase and to explain its slow dissociation. Gastric gland and parietal cell binding of vonoprazan was determined radiometrically. Molecular modelling explained the slow dissociation of vonoprazan from the H(+) ,K(+) -ATPase. Vonoprazan binds selectively to the parietal cell, independent of acid secretion. Vonoprazan binds in a luminal vestibule between the surfaces of membrane helices 4, 5 and 6. Exit of the drug to the lumen is hindered by asp137 and asn138 in the loop between TM1 and TM2, which presents an electrostatic barrier to movement of the sulfonyl group of vonoprazan. This may explain its slow dissociation from the H(+) ,K(+) -ATPase and long-lasting inhibition. The binding model provides a template for design of novel potassium-competitive acid blockers.
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
SAFC
HEPES
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%