Merck
  • Identification of chemical constituents in two traditional Chinese medicine formulae by liquid chromatography-mass spectrometry and off-line nuclear magnetic resonance.
Identification of chemical constituents in two traditional Chinese medicine formulae by liquid chromatography-mass spectrometry and off-line nuclear magnetic resonance.
Journal of pharmaceutical and biomedical analysis (2015-09-24)
Shufang Wang, Yunxiang Zhu, Qing Shao, Yi Wang, Xiaohui Fan, Yiyu Cheng
ABSTRACT
There have been increasing works on identification of the chemical constituents in traditional Chinese medicines (TCMs) by liquid chromatography-mass spectrometry (LC-MS). However, isomers cannot be distinguished generally only by MS data, and the structures of unknown compounds cannot be confirmed. In this study, semi-preparative LC guided by LC-MS was used to prepare the isomers in microscale followed by off-line NMR analysis to confirm their structures. This approach was applied to identifying the constituents in two TCM formulae, Zhi-Zi-Gan-Cao-Chi-Tang (ZZGCCT) and Zhi-Zi-Bai-Pi-Tang (ZZBPT). A total number of 119 constituents were identified tentatively or unambiguously by LC-IT-MS, LC-Q-TOF-MS, and NMR. Among them, 20 constituents were characterized unambiguously by comparing with the reference substances. In addition, 21 constituents without reference substances were prepared for the following 1D-NMR and/or 2D-NMR analysis, and their structures were unambiguously identified by MS, 1D-NMR, and 2D-NMR. Two triterpenoid glycosides (compounds 134 and 140) and one flavonoid glycoside (compound 62a or 62b) were showed to be novel compounds. Compounds 125 and 129, as well as 62a,b, were epimers.
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Citric acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Citric acid, BioUltra, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
2-Hydroxybutyric acid sodium salt, 97%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Citric acid, anhydrous, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Citric acid, 99%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Citric acid, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, for preparative HPLC, ≥99.8% (GC)
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Citric acid, meets analytical specification of Ph. Eur., BP, USP, E330, anhydrous, 99.5-100.5% (based on anhydrous substance)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol-d4, 99 atom % D
Sigma-Aldrich
Methanol-d4, ≥99.8 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol-d4, ≥99.8 atom % D, contains 0.1 % (v/v) TMS
Sigma-Aldrich
Methanol-d4, ≥99.8 atom % D
Sigma-Aldrich
Methanol-d4, ≥99.8 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Methanol-d4, "100%", ≥99.96 atom % D, contains 0.03 % (v/v) TMS