Merck
  • Contribution to the food products' analysis: A research and evaluation on the hemolytic effect of some pesticides used in Lebanon.
Contribution to the food products' analysis: A research and evaluation on the hemolytic effect of some pesticides used in Lebanon.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes (2015-09-12)
Josephine Al-Alam, Maurice Millet, Asma Chbani, Ziad Fajloun
ABSTRACT
Pesticides are a real concern for the society as their use has become critical, leading sometimes to their accumulation as residues in fruits and vegetables. After examining the pesticides sold in Northern Lebanon, this study is focused on the analysis and identification of pesticides residues in fruits and vegetables that are harvested in this region and treated with the locally sold pesticides. Results show: first, (i) a use of Zineb by the name of another pesticide Micronized Sulfur to avoid prosecution; (ii) a significant presence of Metalaxyl in lemons and oranges; (iii) a significant presence of Trifluralin in strawberries; and (iv) a significant presence of Zineb in lemons and tomatoes. Second, with the use of hemolytic tests on human blood results show: (i) a critical concentration and a significant hemolytic effect of some pesticides used in Lebanon; and (ii) an absence of hemolytic effect in the collected fractions of the different analyzed fruit extracts containing pesticides. Finally, this work is the first step for pesticides' analysis in vegetables and fruits in Lebanon, initiating a wider analytical study in order to control and examine the use of pesticides which, according to our results, could have an adverse effect on human health over a long term.
Sigma-Aldrich
Sodium phosphate monobasic, anhydrous, free-flowing, Redi-Dri, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%, free-flowing, Redi-Dri
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Sodium phosphate monobasic solution, BioUltra, 5 M in H2O
Sigma-Aldrich
Sodium phosphate monobasic, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium phosphate dibasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, for molecular biology, ≥98.5% (titration)
Sigma-Aldrich
Sodium phosphate monobasic, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0% (titration)
Sigma-Aldrich
Sodium phosphate dibasic, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Ethyl acetate, natural, ≥99%, FCC, FG
Sigma-Aldrich
Sodium phosphate dibasic, 99.95% trace metals basis
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Copper(II) acetate, 98%
Sigma-Aldrich
Copper(II) acetate, powder, 99.99% trace metals basis
Sigma-Aldrich
Ethyl acetate, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, BioReagent, for molecular biology, anhydrous, ≥98%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis