Merck
  • A new application of cell-free bone regeneration: immobilizing stem cells from human exfoliated deciduous teeth-conditioned medium onto titanium implants using atmospheric pressure plasma treatment.
A new application of cell-free bone regeneration: immobilizing stem cells from human exfoliated deciduous teeth-conditioned medium onto titanium implants using atmospheric pressure plasma treatment.
Stem cell research & therapy (2015-06-20)
Masahiro Omori, Shuhei Tsuchiya, Kenji Hara, Kensuke Kuroda, Hideharu Hibi, Masazumi Okido, Minoru Ueda
ABSTRACT
Surface modification of titanium (Ti) implants promotes bone formation and shortens the osseointegration period. The aim of this study was to promote bone regeneration and stability around implants using atmospheric pressure plasma (APP) pretreatment. This was followed by immobilization of stem cells from human exfoliated deciduous teeth-conditioned medium (SHED-CM) on the Ti implant surface. Ti samples (implants, discs, powder) were treated with APP for 30 seconds. Subsequently, these were immobilized on the treated Ti surface, soaked and agitated in phosphate-buffered saline or SHED-CM for 24 hours at 37 °C. The surface topography of the Ti implants was observed using scanning electron microscopy with energy dispersive X-ray spectroscopy. In vivo experiments using Ti implants placed on canine femur bone were then conducted to permit histological analysis at the bone-implant boundary. For the in vitro experiments, protein assays (SDS-PAGE, Bradford assay, liquid chromatography-ion trap mass spectrometry) and canine bone marrow stromal cell (cBMSC) attachment assays were performed using Ti discs or powder. In the in vitro study, treatment of Ti implant surfaces with SHED-CM led to calcium phosphate and extracellular matrix protein immobilization. APP pretreatment increased the amount of SHED-CM immobilized on Ti powder, and contributed to increased cBMSC attachment on Ti discs. In the in vivo study, histological analysis revealed that the Ti implants treated with APP and SHED-CM stimulated new bone formation around implants. Implant device APP pretreatment followed by SHED-CM immobilization may be an effective application to facilitate bone regeneration around dental implants.
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Guanidine hydrochloride solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Guanidine hydrochloride solution, Colorless liquid, 7.8 - 8.3 M, pH- 4.5 - 5.5
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Guanidine hydrochloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Guanidine hydrochloride, ≥99.0% (AT)
Sigma-Aldrich
Azithromycin
Sigma-Aldrich
Iodoacetamide, ≥99% (NMR), crystalline
Sigma-Aldrich
Guanidine hydrochloride, for molecular biology, ≥99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Urea, BioUltra, for molecular biology, 99% (T)
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture