Merck
  • Effect of Novel Starter Culture on Reduction of Biogenic Amines, Quality Improvement, and Sensory Properties of Doenjang, a Traditional Korean Soybean Fermented Sauce Variety.
Effect of Novel Starter Culture on Reduction of Biogenic Amines, Quality Improvement, and Sensory Properties of Doenjang, a Traditional Korean Soybean Fermented Sauce Variety.
Journal of food science (2015-07-07)
Shruti Shukla, Jong Suk Lee, Hae-Kyong Park, Jung-Ah Yoo, Sung-Yong Hong, Jong-Kyu Kim, Myunghee Kim
ABSTRACT
To select appropriate microorganisms as starter cultures for the reliable and reproducible fermentation of soybean fermented products of Korean Doenjang, various ratios of fungi (Aspergillus oryzae J, Mucor racemosus 15, M. racemosus 42) combined with Bacillus subtilis TKSP 24 were selected as either single, double, or multiple Meju strains for commercial mass production of Doenjang, followed by analysis of sensory characteristics. In the sensory evaluation, Doenjang BAM15-1 and BAM42-1, which were fermented with multiple strains (1:1:1), showed the highest sensory scores as compared to control. Based on sensory characteristics, 6 Doenjang samples were subjected to quantitative determination of amino acids, free sugars, and organic acids (volatile and nonvolatile) contents, followed by determination of biogenic amines. Total sweet taste amino acid contents were highest in BAM15-1 and BAM42-1 samples (333.7 and 295.8 mg/100 g, respectively) and similar that of control (391.1 mg/100 g). Samples BAM15-1 and BAM42-1 showed the relatively high volatile and nonvolatile organic acid contents (154.24, 192.26, and 71.31, 82.42 mg/100 g, respectively). In addition, BAM15-1 and BAM42-1 showed negligible biogenic amine formation, ranging from 0.00 to 1.02 and 0.00 to 3.92 mg/100 g, respectively. These findings indicate that determination of food components along with sensory and quality attributes using multiple microbial Meju strains as a starter culture may provide substantial results on improved quality fermented Doenjang products.
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Spermidine trihydrochloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Perchloric acid, 70%, 99.999% trace metals basis
Sigma-Aldrich
Agmatine sulfate salt, ≥97%, powder
Sigma-Aldrich
Spermidine trihydrochloride, ≥98% (TLC)
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
N-Benzylmethylamine, 97%
Sigma-Aldrich
Dansyl chloride, suitable for fluorescence, BioReagent, ≥99.0% (HPLC)
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Spermine tetrahydrochloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Spermine tetrahydrochloride, ≥99.0% (AT), powder or crystals
Sigma-Aldrich
Phenethylamine, 99%
Sigma-Aldrich
Phenethylamine, purified by redistillation, ≥99.5%
Sigma-Aldrich
Tryptamine hydrochloride, 99%
Sigma-Aldrich
Phenethylamine, ≥99%
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Cadaverine dihydrochloride, ~98%
Sigma-Aldrich
Spermine tetrahydrochloride, BioReagent, for molecular biology
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Putrescine dihydrochloride, ≥98% (TLC)