Merck
  • Phototoxic Risk Assessments on Benzophenone Derivatives: Photobiochemical Assessments and Dermal Cassette-Dosing Pharmacokinetic Study.
Phototoxic Risk Assessments on Benzophenone Derivatives: Photobiochemical Assessments and Dermal Cassette-Dosing Pharmacokinetic Study.
The Journal of pharmacology and experimental therapeutics (2015-05-29)
Yoshiki Seto, Hiroto Ohtake, Masashi Kato, Satomi Onoue
ABSTRACT
This study aimed to qualify photosafety screening on the basis of photochemical and pharmacokinetic (PK) data on dermally applied chemicals. Six benzophenone derivatives (BZPs) were selected as model compounds, and in vitro photochemical/phototoxic characterization and dermal cassette-dosing PK study were carried out. For comparison, an in vivo phototoxicity test was also conducted. All of the BZPs exhibited strong UVA/UVB absorption with molar extinction coefficients of over 2000 M(-1) × cm(-1), and benzophenone and ketoprofen exhibited significant reactive oxygen species (ROS) generation upon exposure to simulated sunlight (about 2.0 mW/cm(2)); however, ROS generation from sulisobenzone and dioxybenzone was negligible. To verify in vitro phototoxicity, a 3T3 neutral red uptake phototoxicity test was carried out, and benzophenone and ketoprofen were categorized to be phototoxic chemicals. The dermal PK parameters of ketoprofen were indicative of the highest dermal distribution of all BZPs tested. On the basis of its in vitro photochemical/phototoxic and PK data, ketoprofen was deduced to be highly phototoxic. The rank of predicted phototoxic risk of BZPs on the basis of the proposed screening strategy was almost in agreement with the results from the in vivo phototoxicity test. The combined use of photochemical and cassette-dosing PK data would provide reliable predictions of phototoxic risk for candidates with high productivity.
Sigma-Aldrich
Propylene glycol, ≥99.5% (GC), FCC, FG
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Sodium phosphate monobasic dihydrate, purum p.a., crystallized, ≥99.0% (T)
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Propylene Glycol, meets USP testing specifications
Sigma-Aldrich
Erythromycin, BioReagent, suitable for cell culture
Sigma-Aldrich
Erythromycin, meets USP testing specifications
Sigma-Aldrich
Imidazole, ≥99% (titration), crystalline
Sigma-Aldrich
Imidazole, ACS reagent, ≥99% (titration)
Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration)
Sigma-Aldrich
Imidazole, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Imidazole, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Imidazole, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Sodium phosphate monobasic dihydrate, BioUltra, for molecular biology, ≥99.0% (T)
Sigma-Aldrich
Erythromycin, potency: ≥850 μg per mg
Sigma-Aldrich
Erythromycin standard solution, 1 mg/mL in H2O
Sigma-Aldrich
Imidazole, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration), free-flowing, Redi-Dri
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%, Redi-Dri, free-flowing
Sigma-Aldrich
Quinine, suitable for fluorescence, anhydrous, ≥98.0% (dried material, NT)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Quinine, 90%
Sigma-Aldrich
N,N-Dimethyl-4-nitrosoaniline, ≥97%
Sigma-Aldrich
1,2-Propanediol, ReagentPlus®, 99%
Sigma-Aldrich
1,2-Propanediol, puriss. p.a., ACS reagent, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Imidazole buffer Solution, BioUltra, 1 M in H2O