Merck
  • Effects of Salt on the Stability of a G-Quadruplex from the Human c-MYC Promoter.
Effects of Salt on the Stability of a G-Quadruplex from the Human c-MYC Promoter.
Biochemistry (2015-05-20)
Byul G Kim, Heather M Evans, David N Dubins, Tigran V Chalikian
ABSTRACT
In an atmosphere of potassium ions, a modified c-MYC NHE III1 sequence with two G-to-T mutations (MYC22-G14T/G23T) forms a highly stable parallel-stranded G-quadruplex. The G-quadruplex exhibits a steady increase in its melting temperature, T(M), with an increase in the concentration of the stabilizing cation K(+). On the other hand, an increase in the concentration of nonstabilizing Cs(+) or TMA(+) cations at a constant concentration of K(+) causes a sharp decline in T(M) followed by a leveling off at ∼200 mM Cs(+) or TMA(+). At 51 °C and 600 μM K(+), an increase in Cs(+) concentration from 0 to 800 mM leads to a complete unfolding of the G-quadruplex. These observations are consistent with the picture in which more counterions accumulate in the vicinity of the unfolded state of MYC22-G14T/G23T (nonspecific ion binding) than in that of the G-quadruplex state. We estimate that the unfolded state condenses one extra counterion compared to the G-quadruplex state. Taken together with our earlier results, our data suggest that sodium or potassium cations sequestered inside the central cavity stabilize the G-quadruplex conformation acting as specifically bound ligands. Nonspecifically bound (condensed) counterions may slightly stabilize, exert no influence (human telomeric G-quadruplexes), or strongly destabilize (MYC22-G14T/G23T) the G-quadruplex conformation. We offer a structural rationalization for the enhanced thermal stability of the MYC22-G14T/G23T G-quadruplex.
Sigma-Aldrich
Phosphoric acid-16O4 solution, 70 wt. % in D2O, 99.9 atom % 16O
Sigma-Aldrich
Phosphoric acid, BioUltra, ≥85% (T)
Sigma-Aldrich
Cesium chloride, ≥99.999% trace metals basis
Sigma-Aldrich
Phosphoric acid, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Cesium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Cesium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Cesium chloride, 99.99% trace metals basis
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Cesium chloride, BioXtra, ≥99.5% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Cesium chloride, for molecular biology, ≥98%
Sigma-Aldrich
Cesium chloride, Grade I, ≥99.0%
Sigma-Aldrich
Cesium chloride, for molecular biology, ≥99% (silver nitrate titration)
Sigma-Aldrich
Cesium chloride, Grade II, ≥98%
Sigma-Aldrich
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Cesium chloride, optical grade, ≥99.5% trace metals basis
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium chloride, for molecular biology, ≥99.0%
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Potassium chloride solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG