Skip to Content
Merck
All Photos(3)

Key Documents

C6914

Sigma-Aldrich

Cesium chloride

Grade II, ≥98%

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
$1,210.00
1 KG
$2,120.00

$1,210.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

Select a Size

Change View
500 G
$1,210.00
1 KG
$2,120.00

About This Item

Linear Formula:
CsCl
CAS Number:
Molecular Weight:
168.36
EC Number:
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:
NACRES:
NA.75
grade:
for analytical purposes
form:
powder
solubility:
H2O: 1 g/mL, clear, colorless

$1,210.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

grade

for analytical purposes

type

Grade II

Assay

≥98%

form

powder

mp

645 °C (lit.)

solubility

H2O: 1 g/mL, clear, colorless

density

3.988 g/cm3

absorption

≤0.2 at 280 in H2O at 50%

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Cl-].[Cs+]

InChI

1S/ClH.Cs/h1H;/q;+1/p-1

InChI key

AIYUHDOJVYHVIT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Used for the preparation of electrically conducting glasses.Used to make solutions for the separation of RNA from DNA by density gradient centrifugation.

Biochem/physiol Actions

Cesium chloride (CsCl) is used to treat various kinds of cancers.[1] Oral and intravenous administration of CsCl is linked with seizures, cardiotoxicity, syncope and death.[1] In rats, CsCl reactions displays several variations in blood chemistry, that are categorized by a reduction of total protein content, pH, an increase in the level of urea, creatinine, glucose and total hemoglobin.[2]

Quality

Solutions may contain slight haze.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biochemical changes in rats under the influence of cesium chloride
Melnykova NM, et al.
Ukrainian Biochemical Journal, 85(2), 105-109 (2013)
Fatal cesium chloride toxicity after alternative cancer treatment
Sessions, et al.
Journal of Alternative and Complementary Medicine (New York, N.Y.), 19(12), 973-975 (2013)
Tuan A Ho et al.
Langmuir : the ACS journal of surfaces and colloids, 28(2), 1256-1266 (2011-12-14)
All-atom molecular dynamics simulations were conducted to study the dynamics of aqueous electrolyte solutions confined in slit-shaped silica nanopores of various degrees of protonation. Five degrees of protonation were prepared by randomly removing surface hydrogen atoms from fully protonated crystalline
Raphaele Germi et al.
The Journal of general virology, 93(Pt 8), 1769-1773 (2012-05-18)
Epstein-Barr virus (EBV), a gammaherpesvirus, infects >90 % of the world's population. Primary infection by EBV can lead to infectious mononucleosis, and EBV persistence is associated with several malignancies. Despite its importance for human health, little structural information is available on
Travis A Hage et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 32(8), 2714-2721 (2012-02-24)
We report a novel coupled system of sodium-activated potassium currents (I(KNa)) and persistent sodium currents (I(NaP)), the components of which are widely distributed throughout the brain. Its existence and importance has not been previously recognized. Although I(KNa) was known to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service