Merck
  • A New Hyaluronic Acid Derivative Obtained from Atom Transfer Radical Polymerization as a siRNA Vector for CD44 Receptor Tumor Targeting.
A New Hyaluronic Acid Derivative Obtained from Atom Transfer Radical Polymerization as a siRNA Vector for CD44 Receptor Tumor Targeting.
Macromolecular bioscience (2015-07-03)
Fabio Salvatore Palumbo, Antonella Bavuso Volpe, Flavia Bongiovì, Giovanna Pitarresi, Gaetano Giammona
ABSTRACT
Two derivatives of hyaluronic acid (HA) have been synthesized by atom transfer radical polymerization (ATRP), starting from an ethylenediamino HA derivative (HA-EDA) and by using diethylaminoethyl methacrylate (DEAEMA) as a monomer for polymerization. Both samples, indicated as HA-EDA-pDEAEMA a and b, are able to condense siRNA, as determined by gel retardation assay and resulting complexes show a size and a zeta potential value dependent on polymerization number, as determined by dynamic light scattering measurements. In vitro studies performed on HCT 116 cell line, that over express CD44 receptor, demonstrate a receptor mediated uptake of complexes, regardless of their surface charge.
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, 53.5-56.5% in H2O
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, 1.0 M in methanol
Sigma-Aldrich
Copper(II) bromide, 99.999% trace metals basis
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, 40 wt. % in H2O
Sigma-Aldrich
Copper(II) bromide, 99%
Sigma-Aldrich
2-Bromo-2-methylpropionic acid, 98%
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
1,2-Dichloroethane, anhydrous, 99.8%
Sigma-Aldrich
Bis(4-nitrophenyl) carbonate, ≥99%
Sigma-Aldrich
Deuterium oxide, 99.8 atom % D
Sigma-Aldrich
Ethylenediamine, ReagentPlus®, ≥99%
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.05 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Diethylamine, ≥99.5%
Sigma-Aldrich
Ethylenediamine solution, technical, 75-80%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 1 % (w/w) 3-(trimethylsilyl)-1-propanesulfonic acid, sodium salt (DSS)
Sigma-Aldrich
Diethylamine, purified by redistillation, 99.5%
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.75 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Sigma-Aldrich
Ethylenediamine, purified by redistillation, ≥99.5%