Merck
  • Investigating the Effect of Cold Soak Duration on Phenolic Extraction during Cabernet Sauvignon Fermentation.
Investigating the Effect of Cold Soak Duration on Phenolic Extraction during Cabernet Sauvignon Fermentation.
Molecules (Basel, Switzerland) (2015-05-07)
Siriwan Panprivech, Larry A Lerno, Charles A Brenneman, David E Block, Anita Oberholster
ABSTRACT
The impact of increasing cold soak (CS) duration (0, 1, 4, 7, and 10 days at 10 °C) on the extraction of phenolic compounds during the CS period and primary fermentation as well as the final composition of Cabernet Sauvignon wine was investigated. The results showed that CS duration had no effect on hydroxycinnamate and flavonol extractions. Greater amounts of gallic acid, (+)-catechin, (-)-epicatechin, and total tannins were extracted with increasing CS duration, with differences maintained during bottle aging. Anthocyanin extraction and color density increased with longer periods of CS; however, by the end of primary fermentation, as well as three months' bottle aging, there were no significant differences due to CS duration. The wines made with seven and 10 days of CS had higher seed tannin contributions and total tannin compared to the non-CS wine, which could potentially result in increased astringency.
Sigma-Aldrich
trans-Ferulic acid, ≥99%
Sigma-Aldrich
Caffeic acid, ≥98.0% (HPLC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, Grade I, ≥99% (GC), suitable for plant cell culture
Sigma-Aldrich
Sucrose, BioUltra, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
trans-Ferulic acid, 99%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Gallic acid monohydrate, ACS reagent, ≥98.0%
Sigma-Aldrich
Gallic acid monohydrate, ≥99% (HPLC)
Sigma-Aldrich
p-Coumaric acid, ≥98.0% (HPLC)
Sigma-Aldrich
(−)-Epicatechin, ≥90% (HPLC)
Sigma-Aldrich
(−)-Epicatechin, ≥98% (HPLC), from green tea
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid