Merck
  • Ammonium increases Ca(2+) signalling and upregulates expression of Cav1.2 gene in astrocytes in primary cultures and in the in vivo brain.
Ammonium increases Ca(2+) signalling and upregulates expression of Cav1.2 gene in astrocytes in primary cultures and in the in vivo brain.
Acta physiologica (Oxford, England) (2015-04-08)
F Wang, T Du, C Liang, A Verkhratsky, L Peng
ABSTRACT
The primary aim of this study was to identify the effects of hyperammonaemia on functional expression of Cav1.2 L-type Ca(2+) channels in astroglia. Primary cultures of mouse astrocytes were used to study effects of chronic treatment (1-5 days) with ammonium chloride, at 1, 3 and 5 mm on depolarization-induced increases in free cytosolic Ca(2+) concentration ([Ca(2+)]i , measured with Fura-2 based microfluorimetry) in control conditions and following treatment with the L-type Ca(2+) channel inhibitor, nifedipine, or with ryanodine receptor inhibitor, ryanodine. Expression of Cav1.2 mRNA was identified with RT-PCR, whereas protein content was determined by Western blotting. Sustained hyperammonaemia in vivo was induced by daily injections of urease (33 units kg body weight(-1), i.p.) for 3 days. Depolarization-induced [Ca(2+)]i transients sensitive to nifedipine (peak of the response) and to ryanodine (plateau phase) were significantly increased in astrocytes chronically exposed to ammonium. The ammonium-induced increase in Ca(2+) influx in astrocytes resulted from an upregulation of Cav1.2 channel's expression detected at mRNA and protein levels. Increase in Cav1.2 expression was prevented by ouabain antagonist canrenone. Similar upregulation of Cav1.2 gene expression was found in the brains of adult mice subjected to intraperitoneal injection of urease. In transgenic mice tagged with an astrocyte-specific or neurone-specific markers and treated with intraperitoneal injections of urease, the fluorescence-activated cell sorting of neurones and astrocytes demonstrated that Cav1.2 mRNA expression was upregulated in astrocytes, but not in neurones. Ammonium-induced deregulation of astroglial Ca(2+) signalling, is, in part, associated with upregulation of Cav1.2 L-type calcium channels.
Sigma-Aldrich
MISSION® esiRNA, targeting human CACNA1C (1)
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
MISSION® esiRNA, targeting human CACNA1C (2)
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Cacna1c
Sigma-Aldrich
DNQX, ≥98% (TLC)
Sigma-Aldrich
Ethidium bromide, ~95% (HPLC)
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate tris salt, ≥97% (HPLC), powder
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
Ethidium bromide, BioReagent, for molecular biology, powder
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Supelco
Pyridaben, PESTANAL®, analytical standard
Sigma-Aldrich
Ethidium bromide solution, for fluorescence, ~1% in H2O
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 10 mg/mL in H2O
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 500 μg/mL in H2O
Sigma-Aldrich
Nifedipine, ≥98% (HPLC), powder
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O