Merck
  • Functionalization of graphene with self-doped conducting polypyrrole by click coupling.
Functionalization of graphene with self-doped conducting polypyrrole by click coupling.
Journal of colloid and interface science (2015-06-10)
Madeshwaran Sekkarapatti Ramasamy, Sibdas Singha Mahapatra, Jae Whan Cho
ABSTRACT
The synthesis of self-doped conducting polypyrrole-grafted graphene sheets (GS-PPy) for non-volatile memory applications is reported. First, the alkyne-modified graphene sheets (GS-alkyne) were covalently functionalized with a water-soluble polymer containing numerous anionic SO3(-) dopants by a copper-catalyzed click reaction. Then, polypyrrole was covalently grafted onto the functionalized graphene sheets by chemical oxidative polymerization to produce GS-PPy hybrids. The GS-PPy hybrids showed a uniform coating of PPy on the GS sheets, good dispersion in aqueous solutions, high electrical conductivity, and red-shifted absorption peak in the UV/Visible spectra. The non-volatile memory device composed of a Al/(GS-PPy/poly(vinyl alcohol))/Al structure, produced by spin coating of the aqueous GS-PPy/poly(vinyl alcohol) solution, showed a good write-once read-many times memory behavior, which was due to good electrical and optical absorption properties of the GS-PPy hybrids. The findings of this study provide a potential solution for the fabrication of water-soluble graphene-based hybrids for non-volatile resistive-memory-based applications.
Sigma-Aldrich
(+)-Sodium L-ascorbate, BioXtra, ≥99.0% (NT)
Sigma-Aldrich
Triethylamine, for protein sequence analysis, ampule, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Sodium hydride, dry, 90%
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
Propargyl alcohol, 99%
Supelco
Tetrahydrofuran, HPLC grade, ≥99.9%, inhibitor-free
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
Pyrrole, ≥98%, FCC, FG
Sigma-Aldrich
Pyrrole, reagent grade, 98%
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Adenosine 5′-phosphosulfate sodium salt, ≥85%
Sigma-Aldrich
(+)-Sodium L-ascorbate, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
(+)-Sodium L-ascorbate, crystalline, ≥98%
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Sodium hydride, 60 % dispersion in mineral oil
Sigma-Aldrich
Thionyl chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Propionitrile, 99%
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
4-Vinylbenzyl chloride, 90%
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
Ammonium persulfate, BioUltra, for molecular biology, ≥98.0% (RT)
Sigma-Aldrich
Ammonium persulfate, BioXtra, ≥98.0%
Sigma-Aldrich
Ammonium persulfate, for molecular biology, suitable for electrophoresis, ≥98%