Merck
  • Aberrant Expression of Dynein light chain 1 (DYNLT1) is Associated with Human Male Factor Infertility.
Aberrant Expression of Dynein light chain 1 (DYNLT1) is Associated with Human Male Factor Infertility.
Molecular & cellular proteomics : MCP (2015-10-04)
Sivankutty Indu, Sreeja C Sekhar, Jeeva Sengottaiyan, Anil Kumar, Sathy M Pillai, Malini Laloraya, Pradeep G Kumar
ABSTRACT
DYNLT1 is a member of a gene family identified within the t-complex of the mouse, which has been linked with male germ cell development and function in the mouse and the fly. Though defects in the expression of this gene are associated with male sterility in both these models, there has been no study examining its association with spermatogenic defects in human males. In this study, we evaluated the levels of DYNLT1 and its expression product in the germ cells of fertile human males and males suffering from spermatogenic defects. We screened fertile (n = 14), asthenozoospermic (n = 15), oligozoospermic (n = 20) and teratozoospermic (n = 23) males using PCR and Western blot analysis. Semiquantitative PCR indicated either undetectable or significantly lower levels of expression of DYNLT1 in the germ cells from several patients from across the three infertility syndrome groups, when compared with that of fertile controls. DYNLT1 was localized on head, mid-piece, and tail segments of spermatozoa from fertile males. Spermatozoa from infertile males presented either a total absence of DYNLT1 or its absence in the tail region. Majority of the infertile individuals showed negligible levels of localization of DYNLT1 on the spermatozoa. Overexpression of DYNLT1 in GC1-spg cell line resulted in the up-regulation of several cytoskeletal proteins and molecular chaperones involved in cell cycle regulation. Defective expression of DYNLT1 was associated with male factor infertility syndromes in our study population. Proteome level changes in GC1-spg cells overexpressing DYNLT1 were suggestive of its possible function in germ cell development. We have discussed the implications of these observations in the light of the known functions of DYNLT1, which included protein trafficking, membrane vesiculation, cell cycle regulation, and stem cell differentiation.
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium orthovanadate, 99.98% trace metals basis
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 10 mg/mL in H2O
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Ethidium bromide solution, for fluorescence, ~1% in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
SAFC
HEPES
Sigma-Aldrich
Sodium orthovanadate, ≥90% (titration)
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, for molecular biology, ≥97.0%
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 500 μg/mL in H2O
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O